Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

HPA017067

Sigma-Aldrich

Anti-NR5A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-α-1-Fetoprotein transcription factor antibody produced in rabbit, Anti-B1-binding factor antibody produced in rabbit, Anti-CYP7A promoter-binding factor antibody produced in rabbit, Anti-Hepatocytic transcription factor antibody produced in rabbit, Anti-LRH-1 antibody produced in rabbit, Anti-Liver receptor homolog 1 antibody produced in rabbit, Anti-Orphan nuclear receptor NR5A2 antibody produced in rabbit, Anti-hB1F antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RQVVHGKEGSIFLVTGQQVDYSIIASQAGATLNNLMSHAQELVAKLRSLQFDQREFVCLKFLVLFSLDVKNLENFQLVEGVQEQVNAALLDYTMCNYPQQTEKFGQLLLRLPEIRAISMQAEEYLYYKHLNGDVPYNNL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR5A2(2494)

Looking for similar products? Visit Product Comparison Guide

General description

NR5A2 (nuclear receptor subfamily 5, group A, member 2) is a member of nuclear receptor subfamily that function as regulatory transcription factors. It is expressed in adult pancreas, preadipocytes, adrenal, sex glands liver, and small intestine. Its expression has also been found in breast adipose tissue and ovary.

Immunogen

Nuclear receptor subfamily 5 group A member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NR5A2 (nuclear receptor subfamily 5, group A, member 2) is associated with the bile acid metabolism, reverse cholesterol transport and cell cycle transitions. It maintains pluripotency in undifferentiated embryonic stem cells by controlling Oct4 expression. It also plays a key role in sustaining estrogen biosynthesis and signaling in ER+ (estrogen receptor) breast cancer. In postmenopausal women, this protein regulates the expression of genes in osteoblasts, and thus, might be linked to osteoporosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70747

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Chien Dominic Heng et al.
Cell stem cell, 6(2), 167-174 (2010-01-26)
Somatic cells can be reprogrammed to induced pluripotent stem cells (iPSCs) with the introduction of Oct4, Sox2, Klf4, and c-Myc. Among these four factors, Oct4 is critical in inducing pluripotency because no transcription factor can substitute for Oct4, whereas Sox2
José A Riancho et al.
European journal of endocrinology, 166(1), 69-75 (2011-11-04)
There is growing evidence for a link between energy and bone metabolism. The nuclear receptor subfamily 5 member A2 (NR5A2) is involved in lipid metabolism and modulates the expression of estrogen-related genes in some tissues. The objective of this study
Cindy Benod et al.
The Journal of biological chemistry, 288(27), 19830-19844 (2013-05-15)
Liver receptor homolog 1 (nuclear receptor LRH-1, NR5A2) is an essential regulator of gene transcription, critical for maintenance of cell pluripotency in early development and imperative for the proper functions of the liver, pancreas, and intestines during the adult life.
Isaac H Solomon et al.
Journal of molecular biology, 354(5), 1091-1102 (2005-11-18)
The DNA-binding and ligand-binding functions of nuclear receptors are localized to independent domains separated by a flexible hinge. The DNA-binding domain (DBD) of the human liver receptor homologue-1 (hLRH-1), which controls genes central to development and metabolic homeostasis, interacts with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service