Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA017000

Sigma-Aldrich

Anti-UTS2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-U-II, Anti-UII, Anti-Urotensin-2 precursor, Anti-Urotensin-II

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

TNVFHLMLCVTSARTHKSTSLCFGHFNSYPSLPLIHDLLLEISFQLSAPHEDARLTPEELERASLLQILPEMLGAERGDILRKADSSTNIFNPRGNLRKFQDFSGQDPNILLSHLLARIWKPYKKRETPDCFWKYCV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UTS2(10911)

General description

UTS2 (urotensin 2) is a vasoactive molecule with a cyclic structure of 11 amino acids. It is expressed in various tissues of human body.

Immunogen

Urotensin-2 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

UTS2 (urotensin 2) is involved in the cell growth and proliferation of the vascular smooth muscle cell. It acts as an endogenous ligand for the orphan G-protein-coupled receptor (GPR 14). It has been studied that UTS2 associates with the menopausal status, and extra-nodal and lymphatic invasion. Research has proved that UTS2 and its receptors have strong correlation with breast cancers. It has also clinicopathological imppact on the cardiovascular physiology and pathophysiology. Thus, studies show upregulation of UII expression in acute coronary syndrome (ACS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86786

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ozan Balakan et al.
Medical science monitor : international medical journal of experimental and clinical research, 20, 1419-1425 (2014-08-13)
Urotensin II is a vasoactive polypeptide. It is known that some vasoactive polypeptides are produced and secreted by tumor cells, and act as a paracrine growth stimulant. The aim of this study was to examine the relationship between urotensin II
Hamood Al Kindi et al.
Peptides, 55, 151-157 (2014-03-20)
Urotensin II (UII) is a vasoactive peptide with various roles in cardiovascular physiology and pathophysiology. There is an accumulating evidence implicating UII in atherosclerosis and coronary artery disease, making it an important target in acute coronary syndrome (ACS). In this
Vadim Le Joncour et al.
Frontiers in cell and developmental biology, 9, 652544-652544 (2021-05-04)
Glioblastomas (GBMs) are the most common primary brain tumors characterized by strong invasiveness and angiogenesis. GBM cells and microenvironment secrete angiogenic factors and also express chemoattractant G protein-coupled receptors (GPCRs) to their advantage. We investigated the role of the vasoactive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service