Skip to Content
MilliporeSigma
All Photos(2)

Documents

HPA015705

Sigma-Aldrich

Anti-ZNF330 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Nucleolar autoantigen 36, Anti-Nucleolar cysteine-rich protein, Anti-Zinc finger protein 330

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

SCQVLEAETFKCVSCNRLGQHSCLRCKACFCDDHTRSKVFKQEKGKQPPCPKCGHETQETKDLSMSTRSLKFGRQTGGEEGDGASGYDAYWKNLSSDKYGDTSYHD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF330(27309)

Immunogen

Zinc finger protein 330 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Zinc finger protein 330 is a protein encoded by the ZNF330 gene in humans and is mapped to chromosome 4q31.1-->q31.2. It is found to be localized in the nucleolus and mitochondria of mammalian cells, due to its nucleolar localization signal (NoLS) that is highly-conserved. This protein is highly conserved and consists of cystein-rich domain. During apoptosis induction, it is translocated from mitochondria to the cytoplasm and helps in releasing cytochrome c.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72583

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan S de Melo et al.
PloS one, 8(3), e59065-e59065 (2013-03-22)
NOA36/ZNF330 is an evolutionarily well-preserved protein present in the nucleolus and mitochondria of mammalian cells. We have previously reported that the pro-apoptotic activity of this protein is mediated by a characteristic cysteine-rich domain. We now demonstrate that the nucleolar localization
J Bolívar et al.
Cytogenetics and cell genetics, 93(3-4), 234-238 (2001-08-31)
We have recently described a novel zinc finger cDNA, ZNF330, which was immunologically characterized as a new human autoantigen, highly conserved during evolution from nematodes to humans. The protein was found at the nucleolus and the cytoplasm in interphase and
Ivan S de Melo et al.
Biochimica et biophysica acta, 1793(12), 1876-1885 (2009-11-10)
Translocations of regulator proteins from or to the mitochondria are key events in apoptosis regulation. NOA36/ZNF330 is a highly evolutionary conserved protein with a characteristic cystein-rich domain. In this work we address its mitochondrial localization and we demonstrate that a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service