Skip to Content
MilliporeSigma
All Photos(5)

Documents

HPA015234

Sigma-Aldrich

Anti-PREX2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DEP.2, Anti-DEPDC2, Anti-FLJ12987, Anti-P-REX2, Anti-PPP1R129

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

GLGFQIRGFGPSVVHAVGRGTVAAAAGLHPGQCIIKVNGINVSKETHASVIAHVTACRKYRRPTKQDSIQWVYNSIESAQEDLQKSHSKPPGDEAGDAFDCKVEEVIDKFNTMAIIDGKKEHVSLTVDNVHLEYGVVYEYDSTAGIKCNV

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PREX2(80243)

General description

PREX2 (phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2) is a homolog of PREX1, and belongs to the Dbl (Diffuse B-cell lymphoma) family guanine nucleotide exchange factors (GEFs) family. It contains a PH (pleckstrin homology) domain, which is essential for substrate recognition. It also contains DH (Dbl-homology) domain and PDZ domains. This protein has a molecular weight of 183kDa.

Immunogen

Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PREX2 (phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2) interacts with and suppresses PTEN (phosphatase and tensin homolog) protein in breast cancer. Mutations in this gene are linked to human melanomas, and abnormal expression of this gene leads to tumorigenesis in immortal melanocytes. Mutations in this gene are also associated with lung, colon and pancreatic cancer. Thus, mutations are thought to turn PREX2 in an oncogene. It recognizes and interacts with Rac-1 protein, RhoA and Cdc42, and activates them. This, in turn, is activated by PI-3K (Phosphoinositide 3-kinase). It is induced by CXCL9 (Chemokine (C-X-C Motif) Ligand 9), which results in enhanced invasiveness of hepatocellular carcinoma. PREX2 is also up-regulated in gastric cancer (GC), and this expression is suppressed by miR-338-3p, which acts as a tumor suppressor in GC.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72469

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiaochu Fu et al.
Molecular pain, 18, 17448069221076460-17448069221076460 (2022-01-28)
Bone cancer pain (BCP) seriously affects the quality of life; however, due to its complex mechanism, the clinical treatment was unsatisfactory. Recent studies have showed several Rac-specific guanine nucleotide exchange factors (GEFs) that affect development and structure of neuronal processes
Xiaoqin Lan et al.
Journal of molecular histology, 45(6), 689-696 (2014-08-26)
Elevated expression of CXCL9 has been shown to involve in the infiltration of inflammatory cells and liver damage after Hepatitis B virus (HBV) infection. However, whether and by what underlying mechanism does CXCL9 play a role in HBV infection associated
Bo Guo et al.
Molecular cancer research : MCR, 12(3), 313-321 (2014-01-01)
Results from recent studies suggest that aberrant microRNA expression is common in numerous cancers. Although miR-338-3p has been implicated in hepatocellular carcinoma, its role in gastric cancer is unknown. To this end, we report that miR-338-3p is downregulated in both
Raji E Joseph et al.
The Journal of biological chemistry, 280(30), 27508-27512 (2005-05-18)
Dbl family guanine nucleotide exchange factors (GEFs) are characterized by the presence of a catalytic Dbl homology domain followed invariably by a lipid-binding pleckstrin homology (PH) domain. To date, substrate recognition and specificity of this family of GEFs has been
Hans Rosenfeldt et al.
FEBS letters, 572(1-3), 167-171 (2004-08-12)
We have identified an activator of Rac, P-REX2, that is structurally related to the exchange factor PtdIns(3,4,5)-dependent Rac exchanger (P-REX1), but exhibits distinct tissue-specific expression. P-REX2 is spliced into two RNA species, approximately 3.5 and approximately 10 kb in size.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service