Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA012756

Sigma-Aldrich

Anti-PTGER4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PGE receptor, EP4 subtype, Anti-Prostaglandin E2 receptor EP4 subtype, Anti-Prostanoid EP4 receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

AASVASRGHPAASPALPRLSDFRRRRSFRRIAGAEIQMV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTGER4(5734)

General description

Prostaglandin E2 receptor EP4 subtype (PTGER4) is a heptahelical G-protein coupled receptor. It belongs to the seven-transmembrane receptor superfamily. The gene encoding it is located on the chromosome arm 5p.

Immunogen

Prostaglandin E2 receptor EP4 subtype recombinant protein epitope signature tag (PrEST)

Application

Anti-PTGER4 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Prostaglandin E2 receptor EP4 subtype (PTGER4) is involved in anti-inflammatory action by binding of its ligand Prostaglandin E2 in macrophages. This interaction between the ligand and receptor increases intracellular cyclic adenosine monophosphate (cAMP) through a G protein called G-α. cAMP in turn activates protein kinase A (PKA) which phosphorylates other downstream effector proteins like cAMP response element–binding protein (CREB). This pathway occurs during bone formation and other developmental activities. The cytoplasmic domain of PTGER4 also interacts with arrestins and causes receptor internalization. PTGER4 also regulates ovarian carcinoma cell invasion and vascular endothelial growth factor production.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72467

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel prostaglandin E receptor 4-associated protein participates in antiinflammatory signaling.
Takayama K, et al.
Circulation Research, 98(4), 499-504 (2006)
Endothelin-1-induced prostaglandin E2-EP2, EP4 signaling regulates vascular endothelial growth factor production and ovarian carcinoma cell invasion.
Spinella F, et al.
The Journal of Biological Chemistry, 279(45), 46700-46705 (2004)
Kristine Kleivi et al.
Molecular cancer, 6, 2-2 (2007-01-05)
Despite the fact that metastases are the leading cause of colorectal cancer deaths, little is known about the underlying molecular changes in these advanced disease stages. Few have studied the overall gene expression levels in metastases from colorectal carcinomas, and
Anna Buchholz et al.
Cancers, 13(6) (2021-04-04)
New prognostic factors and targeted therapies are urgently needed to improve therapeutic outcomes in vulvar cancer patients and to reduce therapy related morbidity. Previous studies demonstrated the important role of prostaglandin receptors in inflammation and carcinogenesis in a variety of
Matthias Oll et al.
BMC urology, 12, 35-35 (2012-12-12)
Prostaglandins play an important role in ureteral obstruction, but the detailed expression profiles of the prostaglandin receptors (PTGER1, PTGER2, PTGER3, PTGER4, PTGFR) remain unknown in the different parts of the human ureter. The expression pattern of PTGER1, PTGER2, PTGER3, PTGER4

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service