Skip to Content
MilliporeSigma
All Photos(8)

Key Documents

HPA012693

Sigma-Aldrich

Anti-OMG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Oligodendrocyte-myelin glycoprotein precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

LPSGLQENIIHLNLSYNHFTDLHNQLTQYTNLRTLDISNNRLESLPAHLPRSLWNMSAANNNIKLLDKSDTAYQWNLKYLDVSKNMLEKVVLIKNTLRSLEVLNLSSNK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OMG(4974)

General description

The gene encoding oligodendrocyte-myelin glycoprotein (OMG) occupies 2.7kb of genomic DNA. It is a glycosylphosphatidyl-inositol-linked protein which is present on the plasma membrane of oligodendrocytes and neurons. OMG contains four domains - a cysteine-rich motif at the amino terminal, a series of leucine-rich repeats (LRs), a serine/threonine-rich region and a hydrophobic carboxy-terminal region.

Immunogen

Oligodendrocyte-myelin glycoprotein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-OMG antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Oligodendrocyte-myelin glycoprotein (OMG) is involved in cell growth regulation, inhibits neurite outgrowth and prevents axonal regeneration after brain injury. Along with two other proteins, Nogo and myelin-associated glycoprotein (MAG) it binds to the Nogo-66 receptor 1 (NgR1) to inhibit axonal growth.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71570

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eric A Huebner et al.
Results and problems in cell differentiation, 48, 339-351 (2009-07-08)
Axon regeneration in the mature mammalian central nervous system (CNS) is extremely limited after injury. Consequently, functional deficits persist after spinal cord injury (SCI), traumatic brain injury, stroke, and related conditions that involve axonal disconnection. This situation differs from that
Oligodendrocyte myelin glycoprotein does not influence node of ranvier structure or assembly.
Chang, K.J., et al.
The Journal of Neuroscience, 30(43), 14476-14481 (2010)
The gene encoding the oligodendrocyte-myelin glycoprotein is embedded within the neurofibromatosis type 1 gene.
D Viskochil
Molecular and Cellular Biology, 11(2), 906-912 (1991)
Patrick Vourc'h et al.
Journal of neurochemistry, 85(4), 889-897 (2003-04-30)
The oligodendrocyte myelin glycoprotein (OMgp) inhibits neurite outgrowth and axonal regeneration after brain injury, but its normal function remains unknown. Several observations suggest its implication in cell growth regulation. Here we report an analysis of the domain requirement in OMgp
D D Mikol et al.
The Journal of cell biology, 110(2), 471-479 (1990-02-01)
The complete primary structure of the human oligodendrocyte-myelin glycoprotein (OMgp), a glycophospholipid-linked membrane protein of oligodendrocytes and central nervous system myelin, has been determined. The deduced amino acid sequence predicts a polypeptide of 433 amino acids which includes a 17-amino

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service