Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA012059

Sigma-Aldrich

Anti-B3GAT2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Beta-1,3-glucuronyltransferase 2, Anti-Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 2, Anti-GlcAT-D, Anti-GlcAT-S, Anti-Glucuronosyltransferase-S, Anti-UDP-glucuronosyltransferase-S

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

PVGLVGGRRYERPLVENGKVVGWYTGWRADRPFAIDMAGFAVSLQVILSNPKAVFKRRGSQPGMQESDFLKQITTVEELEPKANNCTKVLVWHTRTEKVNLANEPKYHLDTVK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B3GAT2(135152)

Immunogen

beta-1,3-glucuronyltransferase 2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

B3GAT2 (β-1,3-glucuronyltransferase 2) is a type II transmembrane protein consisting of a short N-terminal cytoplasmic tail, a transmembrane domain, a stalk, and a large globular catalytic domain. It encodes an amino acid containing protein, which is essentially involved in the biosynthesis of the HNK-1 carbohydrate. It forms the perineuronal nets (TNR, BCAN, NCAN, HAPLN1, HAPLN2, HAPLN3, and HAPLN4) during HNK -1 pathway. The gene is associated with the risk of schizophrenia (SCZ).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71801

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shinako Kakuda et al.
Glycobiology, 15(2), 203-210 (2004-10-08)
The biosynthesis of HNK-1 carbohydrate is mainly regulated by two glucuronyltransferases (GlcAT-P and GlcAT-S) and a sulfotransferase (HNK-1 ST). To determine how the two glucuronyltransferases are involved in the biosynthesis of the HNK-1 carbohydrate, we prepared soluble forms of GlcAT-P
Irene Marcos et al.
Journal of human genetics, 47(12), 677-680 (2003-01-11)
We report on the structure, map location, and tissue expression of the human GlcAT-Sgene. The gene covers approximately 85 Kb on chromosome 6 (6q13) between the D6S455 and D6S1673 markers. GlcAT-S is composed of four exons and encodes a 324-amino-acid
Anna K Kähler et al.
Biological psychiatry, 69(1), 90-96 (2010-10-19)
The Human Natural Killer-1 carbohydrate (HNK-1) is involved in neurodevelopment and synaptic plasticity. Extracellular matrix structures called perineuronal nets, condensed around subsets of neurons and proximal dendrites during brain maturation, regulate synaptic transmission and plasticity. Ten genes of importance for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service