Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA011126

Sigma-Aldrich

Anti-BTN1A1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BT, Anti-Butyrophilin subfamily 1 member A1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable

immunogen sequence

DYESGDISFYNMNDGSDIYTFSNVTFSGPLRPFFCLWSSGKKPLTICPIADGPERVTVIANAQDLSKEIPLSPMGEDSAPRDADTLHSKLIPTQPSQGA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BTN1A1(696)

General description

BTN1A1 (butyrophilin, subfamily 1, member A1) belongs to the BTN gene family, which resides within the major histocompatibility complex class I region on gene 6p22.1. This protein is composed of 526 amino acids and has a hydrophobic signal sequence at its N-terminal, which is made of 26 amino acids. The signal peptide is cleaved off before the secretion of the protein. BTN1A1 also belongs to immunoglobulin (Ig) superfamily, where the Ig domains are present in the extracellular region. The protein has a transmembrane region and short heptad repeats in the cytoplasmic tail. It is predominantly expressed in the epithelium of lactating mammary gland.

Immunogen

Butyrophilin subfamily 1 member A1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BTN1A1 (butyrophilin, subfamily 1, member A1) is a milk protein which is secreted in milk in the lipid droplets. It controls the secretion of lipid droplets in milk, by interacting with xanthine dehydrogenase/oxidase, and acting either as a receptor or structural protein. Inactivation of this gene in mice leads to the formation of triacylglycerol pools in the cytoplasm. When present in dietary products, BTN1A1 leads to alterations in multiple sclerosis (MS). This is because of the similarity in the IgI domain of BTN1A1 and the IgV domain of myelin oligodendrocyte glycoprotein (MOG). MOG is the antigen for auto-antibodies, and is found on the myelin nerve sheath of MS patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71881

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherry L Ogg et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(27), 10084-10089 (2004-07-01)
Butyrophilin 1a1 (Btn1a1), which is a member of the Ig superfamily, is highly expressed in the lactating mammary gland and is secreted into milk in association with lipid droplets. To determine the potential function of Btn1a1 in milk secretion, we
D A Rhodes et al.
Genomics, 71(3), 351-362 (2001-02-15)
We sequenced the 170-kb cluster of BTN genes in the extended major histocompatibility complex region, 4 Mb telomeric of human leukocyte antigen class I genes, at 6p22.1. The cluster consists of seven genes belonging to the expanding B7/butyrophilin-like group, a
Jaekwang Jeong et al.
The Journal of biological chemistry, 284(33), 22444-22456 (2009-06-18)
Butyrophilin 1A1 (BTN1A1) and xanthine oxidoreductase (XOR) are highly expressed in the lactating mammary gland and are secreted into milk associated with the milk fat globule membrane (MFGM). Ablation of the genes encoding either protein causes severe defects in the
I H Mather et al.
Journal of dairy science, 76(12), 3832-3850 (1993-12-01)
The molecular and cellular biology of the milk protein butyrophilin is reviewed. Butyrophilin constitutes more than 40% by weight of the total protein associated with the fat globule membrane of bovine milk. Closely related proteins are abundant in the fat

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service