Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA010814

Sigma-Aldrich

Anti-DNAJB11 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-DnaJ homolog subfamily B member 11 precursor antibody produced in rabbit, Anti-ER-associated Hsp40 co-chaperone antibody produced in rabbit, Anti-ER-associated dnaJ protein 3 antibody produced in rabbit, Anti-ErJ3 antibody produced in rabbit, Anti-PWP1-interacting protein 4 antibody produced in rabbit, Anti-hDj9 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EPGVRDGMEYPFIGEGEPHVDGEPGDLRFRIKVVKHPIFERRGDDLYTNVTISLVESLVGFEMDITHLDGHKVHISRDKITRPGAKLWKKGEGLPNFDNNNIKGSLIITFDVDFPKEQLTEEAREGIKQLLKQGSVQKVYNGLQ

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DNAJB11(51726)

Looking for similar products? Visit Product Comparison Guide

General description

DNAJB11 (DnaJ (Hsp40) homolog, subfamily B, member 11) belongs to family of endoplasmic reticulum (ER) Hsp40/DnaJ family. It is a part of ERAD (Endoplasmic-reticulum-associated protein degradation), and is a heat shock protein 40 (HSP40) chaperone. It contains the J domain, which is a highly conserved region made of 75 amino acids. It also shares certain domains with other HSP40 chaperones. These domains include a C-terminal domain, a glycine/phenylalanine-rich putative linker region and a cystine-rich domain. It is a soluble glycoprotein, oriented towards the luminal region of endoplasmic reticulum (ER). DNAJB11 is expressed ubiquitously, and has the highest expression in secretory tissues.

Immunogen

DnaJ heat shock protein family (Hsp40) member B11 recombinant protein epitope signature tag (PrEST)

Application

Anti-DNAJB11 antibody is suitable for immunoprecipitation. Anti-DNAJB11 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

DNAJB11 (DnaJ (Hsp40) homolog, subfamily B, member 11) is a part of unassembled immunoglobulin (Ig) heavy chain:BiP (binding immunoglobulin protein) complex, where it acts as a co-factor for BiP, and helps in folding and assembly of proteins. It plays a major role in mammalian cell resistance to vero toxin. It is also involved in ER (endoplasmic reticulum) stress tolerance. It is secreted in the extracellular space, where it binds to mis-folded proteins and prevents their aggregation. It also reduces the toxicity of disease-related prion protein. DNAJB11 is involved in the intoxication pathway of cholera toxin.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71694

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katsuya Nakanishi et al.
Cell stress & chaperones, 9(3), 253-264 (2004-11-17)
Heat shock protein 40 (Hsp40) family proteins are known to bind to Hsp70 through their J-domain and regulate the function of Hsp70 by stimulating its adenosine triphosphatase activity. In the endoplasmic reticulum (ER), there are 5 Hsp40 family proteins known
Steffen Israel et al.
Scientific reports, 9(1), 13356-13356 (2019-09-19)
Early mouse embryos have an atypical translational machinery that consists of cytoplasmic lattices and is poorly competent for translation. Hence, the impact of transcriptomic changes on the operational level of proteins is predicted to be relatively modest. To investigate this
Wen-Li Lu et al.
Evidence-based complementary and alternative medicine : eCAM, 2014, 192749-192749 (2014-11-13)
Akebia Fructus has long been used for hepatocellular carcinoma (HCC) in China, while the molecular mechanism remains obscure. Our recent work found that Akebia trifoliate (Thunb.) Koidz seed extract (ATSE) suppressed proliferation and induced endoplasmic reticulum (ER) stress in SMMC-7721.
M Yu et al.
The Journal of biological chemistry, 275(32), 24984-24992 (2000-05-29)
Hsp40 co-chaperones, characterized by the presence of a highly conserved J domain, are involved in nearly all aspects of protein synthesis, folding, and secretion. Within the lumen of the endoplasmic reticulum, these chaperones are also involved in reverse translocation and
Joseph C Genereux et al.
The EMBO journal, 34(1), 4-19 (2014-11-02)
The Unfolded Protein Response (UPR) indirectly regulates extracellular proteostasis through transcriptional remodeling of endoplasmic reticulum (ER) proteostasis pathways. This remodeling attenuates secretion of misfolded, aggregation-prone proteins during ER stress. Through these activities, the UPR has a critical role in preventing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service