Skip to Content
MilliporeSigma
All Photos(6)

Documents

HPA009139

Sigma-Aldrich

Anti-CDH15 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cadherin-14 antibody produced in rabbit, Anti-Cadherin-15 precursor antibody produced in rabbit, Anti-M-cadherin antibody produced in rabbit, Anti-Muscle-cadherin antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

TFSARDPDTEQLQRLSYSKDYDPEDWLQVDAATGRIQTQHVLSPASPFLKGGWYRAIVLAQDDASQPRTATGTLSIEILEVNDHAPVLAPPPPGSLCSEPHQGPGLLLGATDEDLPPHGAPFHFQLSPRLPELGRNWSLSQVNVSH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH15(1013)

General description

CDH15 (cadherin 15) is a member of the transmembrane glycoprotein adhesion-mediating protein family called cadherins. CDH15 is also known as MCAD or muscle-cadherin. CDH15 mRNA is present in low amount in myoblasts and in high levels in myotube-forming cells. This protein is predominantly expressed in skeletal muscle and brain. This gene is localized to human chromosome 16q, spans 24.3kb and is composed of 14 exons.The encoded protein is composed of 814 amino acids, and is a member of the type I classic cadherin superfamily group. It contains a single transmembrane domain and five extracellular (EC) repeats, which mediate Ca2+-dependent homophilic cell-cell interaction.

Immunogen

Cadherin-15 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CDH15 (cadherin 15) is responsible for hemophilic cell-cell adhesion, and functions as an interacting partner of β-catenin. β-catenin, in turn interacts with α-catenin, which is essential for the control of cytoskeletal function at adherens junctions. Changes in this gene, either independently or in combination with other factors, are linked with intellectual disability (ID). This protein is also responsible for neuronal cell-cell adhesion, and plays a part in neural morphogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71903

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Donalies et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(18), 8024-8028 (1991-09-15)
Cadherins, a multigene family of transmembrane glycoproteins, mediate Ca(2+)-dependent intercellular adhesion. They are thought to be essential for the control of morphogenetic processes, including myogenesis. Here we report the identification and characterization of the cDNA of another member of the
T Shibata et al.
The Journal of biological chemistry, 272(8), 5236-5240 (1997-02-21)
Cadherins, a family of Ca2+-dependent cell-cell adhesion molecules, mediate neural cell-cell interactions and may play important roles in neural development. By searching for molecules that interact with beta-catenin, a cytoplasmic regulator of cadherins, we have identified a new member of
Kavita Bhalla et al.
American journal of human genetics, 83(6), 703-713 (2008-11-18)
Cell-adhesion molecules play critical roles in brain development, as well as maintaining synaptic structure, function, and plasticity. Here we have found the disruption of two genes encoding putative cell-adhesion molecules, CDH15 (cadherin superfamily) and KIRREL3 (immunoglobulin superfamily), by a chromosomal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service