Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

HPA008927

Sigma-Aldrich

Anti-FNDC3A antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fibronectin type-III domain-containing protein 3a antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

GAGDMTTQYMPQYQSSQVYGDVDAHSTHGRSNFRDERSSKTYERLQKKLKDRQGTQKDKMSSPPSSPQKCPSPINEHNGLIKGQIAGGINTGSAKIKSGKGKGGTQVDTEIEEKDEETKAFEALLSNIVKPVA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FNDC3A(22862)

Looking for similar products? Visit Product Comparison Guide

General description

FNDC3A (fibronectin type III domain containing 3A) gene is localized to human chromosome 13q14.2 and is composed of 26 exons and 25 introns. The encoded protein is composed of 1198 amino acids and has a molecular weight of 131.8kDa. This protein is also called HUGO (human gene expressed in odontoblasts) and exists as two isoforms- HUGO1 and HUGO2. This protein contains eight type III fibronectin domains. The N-terminal contains a proline rich region and the C-terminal contains a transmembrane helix. This protein is highly expressed in odontoblast-like cells and trachea, with comparatively less expression in brain, lung, kidney and liver.

Immunogen

Fibronectin type-III domain-containing protein 3a recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Fibronectin type-III domain-containing protein 3a is a protein encoded by the FNDC3A gene in humans. It is also known as HUGO and is mainly composed of a proline-rich region at the N-terminus, 8 type III-fibronectin modules, and a transmembranous helix at the C-terminus. This protein is expressed in nerve fibers in the dental pulp and other tissues. In odontoblasts, it is located in Golgi vesicles and may have a role in collagen and glycosaminoglycan synthesis. Polymorphisms in this gene is associated with Autism spectrum disorder (ASD), a neurodevelopmental syndrome associated with impairments of reciprocal communication and cognitive function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71727

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Liedtke et al.
Scientific reports, 9(1), 13383-13383 (2019-09-19)
Fin development and regeneration are complex biological processes that are highly relevant in teleost fish. They share genetic factors, signaling pathways and cellular properties to coordinate formation of regularly shaped extremities. Especially correct tissue structure defined by extracellular matrix (ECM)
Myungja Ro et al.
Psychiatry research, 209(2), 246-248 (2013-05-04)
Autism spectrum disorder (ASD) is a neurodevelopmental syndrome associated with impairments of reciprocal communication and cognitive function. Associations between single-nucleotide polymorphisms (SNPs) and ASD were analysed by logistic regression. Polymorphisms in fibronectin type III domain-containing 3A (FNDC3A) exhibited significant associations
F Carrouel et al.
Journal of dental research, 87(2), 131-136 (2008-01-26)
Previously, we established a subtractive cDNA library enriched in odontoblast-specific genes and hypothesized that new, previously unidentified, markers would be present, associated with the odontoblast phenotype. In this paper, we report the first characterization of a new gene we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service