Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

HPA007876

Sigma-Aldrich

Anti-MMEL1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

MMEL2, NEPII, NL1, NL2, SEP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$827.00

$827.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$827.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

$827.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMEL1(79258)

Immunogen

Membrane metallo-endopeptidase-like 1 recombinant protein epitope signature tag (PrEST)

Sequence
LEELQSQFGLKGFNWTLFIQTVLSSVKIKLLPDEEVVVYGIPYLQNLENIIDTYSARTIQNYLVWRLVLDRIGSLSQRFKDTRVNYRKALFGTMVEEVRWRECVGYVNSNMENAVGSLYVREAFPGDSKSMVRELIDKVRTVFVETLD

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Membrane metallo-endopeptidase-like 1 (MMEL1) is a type II transmembrane protein that may function in neuropeptide degradation. It also plays a role in the degradation of β-amyloid and is therefore, linked to Alzheimer′s disease. Defects in this gene have been associated with multiple sclerosis. Aberration in this gene has also been linked to primary biliary cirrhosis. Variant of MMEL1 has been associated with rheumatoid arthritis in the Han Chinese population.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70332

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gideon M Hirschfield et al.
Nature genetics, 42(8), 655-657 (2010-07-20)
We genotyped individuals with primary biliary cirrhosis and unaffected controls for suggestive risk loci (genome-wide association P < 1 x 10(-4)) identified in a previous genome-wide association study. Combined analysis of the genome-wide association and replication datasets identified IRF5-TNPO3 (combined
Patrick Danoy et al.
Annals of the rheumatic diseases, 70(10), 1793-1797 (2011-07-26)
The genome-wide association study era has made great progress in identifying susceptibility genes and genetic loci for rheumatoid arthritis (RA) in populations of White European ancestry. However, few studies have tried to dissect disease aetiopathogenesis in other ethnic populations. To
M Ban et al.
Genes and immunity, 11(8), 660-664 (2010-06-25)
Several single-nucleotide polymorphism (SNP) genome-wide association studies (GWASs) have been completed in multiple sclerosis (MS). Follow-up studies of the variants with the most promising rankings, especially when supplemented by informed candidate gene selection, have proven to be extremely successful. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service