Skip to Content
MilliporeSigma
All Photos(4)

Documents

HPA007666

Sigma-Aldrich

Anti-CDK20 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab1

Synonym(s):

Anti-CCRK, Anti-CDK20, Anti-RAB6KIFL, Anti-cyclin-dependent kinase 20, Anti-p42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RSVGCIMGELLNGSPLFPGKNDIEQLCYVLRILGTPNPQVWPELTELPDYNKISFKEQVPMPLEEVLPDVSPQALDLLGQFLLYPPHQRIAASK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCRK(23552)

Immunogen

cyclin-dependent kinase 20 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CDK20 (cyclin-dependent kinase 20) gene encodes a protein kinase that is similar to CDK7, the catalytic subunit of the CDK-activating kinase (CAK) complex. CDKs require association with specific cyclin subunits and are involved in cellular processes such as cell cycle or transcription. The encoded protein phosphorylates Thr-160 and activates CDK2 that is involved in the regulation of cell cycle. It has been identified as a candidate oncogene in human glioblastoma. It is involved in the carcinogenesis of colorectal cancer. It is also called as CCRK (cell cycle-related kinase) and mediates the G1/S transition in cell cycle by regulating Cdk2, cyclin E and Rb.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71439

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Liu et al.
The Journal of biological chemistry, 279(6), 4507-4514 (2003-11-05)
The cyclin-dependent kinase (CDK)-activating kinase (CAK) phosphorylates a conserved threonine residue on CDKs and activates them. Two known classes of CAKs are represented by monomeric Cak1p in budding yeast Saccharomyces cerevisiae and by heterotrimeric CDK7-cyclin H-Mat1 in human and other
Xiaomeng An et al.
European journal of cancer (Oxford, England : 1990), 46(9), 1752-1761 (2010-05-15)
Cell cycle-related kinase (CCRK) is a newly identified protein kinase homologous to Cdk7. We have previously shown that CCRK is a candidate oncogene in human glioblastoma. However, whether CCRK is a bona fide oncogene remains to be tested. The aim

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service