Skip to Content
MilliporeSigma
All Photos(1)

Documents

HPA007526

Sigma-Aldrich

Anti-GREM1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CKTSF1B1, Anti-CRAC1, Anti-DAND2, Anti-DRM, Anti-HMPS, Anti-gremlin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KKGSQGAIPPPDKAQHNDSEQTQSPQQPGSRNRGRGQGRGTAMPGEEVLESSQEALHVTERKYLKRDWCKTQPLKQTIHEEGCNSRTIINRFC

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GREM1(26585)

General description

GREM1 is a protein of DAN family of BMP (bone morphogenic protein) antagonists involved in developmental and transformation related function. It can also interact with signalling EC (endothelial cell) receptors in a BMP- independent manner and can induce in vitro sprouting of EC cells and in vivo angiogenesis. In humans this protein is highly expressed in small intestine, adult and foetal brain and colon . Anti-GREM1 antibody can be used in western blotting. Rabbit anti-GREM1 antibodies are specific for GREM1 protein.

Immunogen

Gremlin-1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71353.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yunjia Wang et al.
BioMed research international, 2020, 4243015-4243015 (2020-10-09)
Adolescent idiopathic scoliosis (AIS) is a common complex disease, and bone homeostasis plays an important role in its pathogenesis. Recent advances in epigenetic research show that dysregulated miRNAs may participate in the development of orthopedic diseases and AIS. The aim
Stefania Mitola et al.
Blood, 112(4), 1154-1157 (2008-05-29)
Recent observations have shown that Drm, a member the Dan family of bone morphogenic protein (BMP) antagonists, induces endothelial cell (EC) sprouting in vitro and angiogenesis in vivo by interacting with signaling EC receptors in a BMP-independent manner. Here, recombinant
L Z Topol et al.
Cytogenetics and cell genetics, 89(1-2), 79-84 (2000-07-15)
We have mapped and characterized the human homolog of Drm/Gremlin (CKTFS1B1), a member of a family of BMP antagonists that have been linked to both developmental and transformation-related functions. By screening a human cDNA library, we isolated a 3.3-kb cDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service