Skip to Content
MilliporeSigma
All Photos(2)

Documents

HPA006686

Sigma-Aldrich

Anti-ZNF396 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger and SCAN domain-containing protein 14 antibody produced in rabbit, Anti-Zinc finger protein 396 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FEKRQGNPSWKKQQKCDECGKIFSQSSALILHQRIHSGKKPYACDECAKAFSRSAILIQHRRTHTDSKYEHAIAEAQKNMYFKTRLKCPRSLSG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF396(252884)

Immunogen

Zinc finger protein 396 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF396 (Zinc finger protein 396) is a novel zinc finger protein expressed highly in the liver. It consists of a SCAN (SRE-ZBP, CTfin51, AW-1, and Number 18 cDNA or ZnF20) domain in the N-terminal region and several consecutive C(2)H(2) zinc finger motifs at the C-terminal end. It possesses a transcriptional repression activity. ZNF396 is expressed in basal cell carcinoma (BCC). It has been reported that ZNF396 may play a role in the Notch-Hes1 (hairy and enhancer of split-1) signaling pathway. It suppresses expression of the notch system effector molecules and prevents BCC cells to enter into the Notch-mediated squamous cell differentiation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71005

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juncheng Bai et al.
Archives of dermatological research, 306(4), 399-404 (2014-01-22)
Zfp191 represses differentiation and keeps various cells in the stem/progenitor stage. Here, we report that a Zfp191 homolog protein, ZNF396, is expressed in basal cell carcinoma (BCC) and possibly represses the expression of a Notch system effector molecule, Hes1 (hairy
Yimin Wu et al.
Gene, 310, 193-201 (2003-06-13)
We have identified two novel human SCAN domain genes ZNF396 and ZNF397, which are clustered within the region of chromosome 18q12. Three isoforms of ZNF396 transcript, 1.5, 2.5 and 3.9-kb, are expressed highly in liver. Four isoforms of ZNF397 transcript

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service