Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA006018

Sigma-Aldrich

Anti-CALU antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Calumenin precursor, Anti-Crocalbin, Anti-IEF SSP 9302

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000
western blot: 0.04-0.4 μg/mL

immunogen sequence

TKEEFTAFLHPEEYDYMKDIVVQETMEDIDKNADGFIDLEEYIGDMYSHDGNTDEPEWVKTEREQFVEFRDKNRDGKMDKEETKDWILPSDYDHAEAEARHLVYESDQNKDGKLTKEEIVDKYD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALU(813)

Immunogen

Calumenin precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CALU antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Calumenin is a protein encoded by the CALU gene in humans and is mapped to chromosome 7q32. It belongs to a family of multiple EF-hand proteins that include the ER localized proteins reticulocalbin and ERC-55, and the golgi localized Cab45. This protein regulates proteins involved in coagulation and arterial calcification, and it also has extracellular functions related to atherosclerosis. It is found to be localized in close association with the ER membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86562

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Hernández-Romero et al.
Annals of medicine, 42(6), 439-446 (2010-08-03)
Arterial calcification is a risk factor for atherosclerosis. Calumenin (CALU), a protein regulating proteins involved in coagulation and arterial calcification also has extracellular functions related to atherosclerosis. We recently described that CALU polymorphism A29809G was related to acenocoumarol requirements, and
Human calumenin gene (CALU): cDNA isolation and chromosomal mapping to 7q32.
D Yabe et al.
Genomics, 49(2), 331-333 (1998-05-23)
H Vorum et al.
Biochimica et biophysica acta, 1386(1), 121-131 (1998-07-24)
By microsequencing and cDNA cloning we have identified the transformation-sensitive protein No. IEF SSP 9302 as the human homologue of calumenin. The nucleotide sequence predicts a 315 amino acid protein with high identity to murine and rat calumenin. The deduced
H Vorum et al.
Experimental cell research, 248(2), 473-481 (1999-05-01)
Calumenin belongs to a family of multiple EF-hand proteins that include reticulocalbin, ERC-55, and Cab45. Reticulocalbin and ERC-55 localize to the ER due to a C-terminal HDEL retrieval signal. Cab45 contains a HEEF C-terminal sequence and is localized to the
M Vasiljevic et al.
Neuroscience, 202, 29-41 (2011-12-20)
Calumenin is a Ca(2+)-binding protein that belongs to the CREC superfamily. It contains six EF-hand domains that exhibit a low affinity for Ca(2+) as well as an endoplasmic reticulum retention signal. Calumenin exhibits a broad and relatively high expression in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service