Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA003607

Sigma-Aldrich

Anti-SERPINA4 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Kallikrein inhibitor antibody produced in rabbit, Anti-Kallistatin precursor antibody produced in rabbit, Anti-Protease inhibitor 4 antibody produced in rabbit, Anti-Serpin A4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$827.00

$827.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
$827.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

$827.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

NLTELSESDVHRGFQHLLHTLNLPGHGLETRVGSALFLSHNLKFLAKFLNDTMAVYEAKLFHTNFYDTVGTIQLINDHVKKETRGKIVDLVSELKKDVLMVLVNYIYFKALWEKPFISSRTTPKDFYVDENTTVRVPMMLQDQE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINA4(5267)

Immunogen

Kallistatin precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Kallistatin is a protein encoded by the SERPINA4 gene in humans and belongs to serpin superfamily. The tissue kallikrein is mainly localized to goblet cells and kallistatin to epithelial cells of human intestine. It binds to tissue kallikrein and inhibits kallikrein′s kininogenase as well as amidolytic activities. It is a a novel tissue kallikrein inhibitor that possesses anti-inflammatory and anti-oxidant properties and may have some therapeutic roles in cardiometabolic disorders. It may also be useful for the treatment of lung cancer due to its anti-angiogenic and anti-inflammatory activities that may be effective in inhibiting tumor metastasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70551

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antoni Stadnicki et al.
International immunopharmacology, 3(7), 939-944 (2003-06-18)
The profile of tissue kallikrein (TK) and its inhibitor, kallistatin was evaluated in patients with active ulcerative colitis (UC) and Crohn's disease (CD). Tissue kallikrein is mainly localized to goblet cells and kallistatin to epithelial cells of human intestine. Intestinal
K X Chai et al.
The Journal of biological chemistry, 268(32), 24498-24505 (1993-11-15)
We have recently purified a novel human serine proteinase inhibitor (serpin), designated as kallistatin, which binds to tissue kallikrein and inhibits kallikrein's kininogenase and amidolytic activities. In the present studies, we have cloned a full-length cDNA encoding kallistatin from human
Haidong Zhu et al.
Metabolism: clinical and experimental, 62(5), 642-646 (2012-11-30)
It is generally recognized that obesity and cardiometabolic risk are more prevalent in African Americans. Kallistatin, a novel tissue kallikrein inhibitor, has anti-inflammatory and anti-oxidant properties. Thus, the goal of this study was to examine the relationships among plasma kallistatin
Ai-Li Shiau et al.
BMC cancer, 10, 245-245 (2010-06-01)
Angiogenesis plays an important role in the development and progression of tumors. Kallistatin exerts anti-angiogenic and anti-inflammatory activities that may be effective in inhibiting tumor metastasis. We investigated the antitumor effect of lentivirus-mediated kallistatin gene transfer in a syngeneic murine

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service