Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA003359

Sigma-Aldrich

Anti-BRPF1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ARPC4, Anti-HOTTL, Anti-Tubulin-tyrosine ligase-like protein 3, AntiBR140 protein, AntiBromodomain and PHD finger-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SLAGDEATHHTEDAAEEERLVLLENQKHLPVEEQLKLLLERLDEVNASKQSVGRSRRAKMIKKEMTALRRKLAHQRETGRDGPERHGPSSRGSLTPHPAACDKDGQTD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRPF1(7862)

General description

BRPF1 (bromodomain and plant homeodomain finger containing, 1) protein contains plant homeodomain (PHD) fingers, a bromodomain that recognizes acetylated histones, and a proline-tryptophan-tryptophan-proline (PWWP) domain.

Immunogen

Peregrin recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

BRPF1 (bromodomain and plant homeodomain finger containing, 1) gene encodes a component of the monocytic leukemic zinc finger (MOZ) histone acetyltransferase (HAT), a quaternary complex that acetylates histones. This component activates MOZ HAT by linking the MOZ catalytic subunit to the ING5 (inhibitor of growth 5) and hEaf6 subunits and promotes acetylation of histones. It also recruits Moz to active chromatin during mitosis. The MOZ HAT complex also functions in proper patterning for skeletal development. The PWWP domain of this protein associates with H3K36me3 (Trimethylation of Lys36 in histone H3) that mediates the elongation phase of transcription and functions as an epigenetic regulator of cell growth and differentiation. The effector domains of human BRPF1 are involved in gene transcription, chromatin binding and remodeling.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84803

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandro Vezzoli et al.
Nature structural & molecular biology, 17(5), 617-619 (2010-04-20)
Trimethylation of Lys36 in histone H3 (H3K36me3) coordinates events associated with the elongation phase of transcription and is also emerging as an important epigenetic regulator of cell growth and differentiation. We have identified the PWWP domain of bromo and plant
Kathrin Laue et al.
Development (Cambridge, England), 135(11), 1935-1946 (2008-05-13)
The Trithorax group (TrxG) is composed of diverse, evolutionary conserved proteins that form chromatin-associated complexes accounting for epigenetic transcriptional memory. However, the molecular mechanisms by which particular loci are marked for reactivation after mitosis are only partially understood. Here, based
Amanda Poplawski et al.
Journal of molecular biology, 426(8), 1661-1676 (2013-12-18)
The monocytic leukemic zinc finger (MOZ) histone acetyltransferase (HAT) acetylates free histones H3, H4, H2A, and H2B in vitro and is associated with up-regulation of gene transcription. The MOZ HAT functions as a quaternary complex with the bromodomain-PHD finger protein
Kenta Hibiya et al.
Developmental biology, 329(2), 176-190 (2009-03-04)
The epigenetic mechanism involving chromatin modification plays a critical role in the maintenance of the expression of Hox genes. Here, we characterize a mutant of the medaka fish, named biaxial symmetries (bis), in which brpf1, a subunit of the MOZ
Mulu Y Lubula et al.
FEBS letters, 588(21), 3844-3854 (2014-10-05)
Bromodomain-PHD finger protein 1 (BRPF1) is part of the MOZ HAT complex and contains a unique combination of domains typically found in chromatin-associated factors, which include plant homeodomain (PHD) fingers, a bromodomain and a proline-tryptophan-tryptophan-proline (PWWP) domain. Bromodomains are conserved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service