Skip to Content
MilliporeSigma
All Photos(2)

Documents

HPA003354

Sigma-Aldrich

Anti-ZNF74 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Zinc finger protein 74 antibody produced in rabbit, Anti-hZNF7 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GEKPFDCSQCWKAFSCHSSLIVHQRIHTGEKPYKCSECGRAFSQNHCLIKHQKIHSGEKSFKCEKCGEMFNWSSHLTEHQRLHSEGKPLAIQFNKHLLSTYYVPGSLLGAGDAGLRDVDPIDALDVAKLLCVVP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF74(7625)

General description

ZNF74 (zinc finger protein 74) gene belongs to the transcription factor IIIA/Kruppel family and contains three exons. Exon 2 encodes an N-terminal truncated Kruppel-associated box (KRAB). Exon 3 encodes the zinc finger domain and the 3′-untranslated region (UTR). The protein encoded by this gene is approx. 64kDa and is localized to the nucleus. It is specifically expressed in neural crest-derived tissues and neuroepithelium of the spinal cord as well as in foregut endoderm epithelia (esophagus and respiratory tract).

Immunogen

Zinc finger protein 74 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF74 (zinc finger protein 74) gene encodes an RNA-binding protein that preferentially binds to poly(U) and poly(G) RNA homopolymers via the zinc finger domain. It is found associated with the nuclear matrix that is involved in the replication of DNA and in RNA synthesis and processing. The protein may be involved in RNA metabolism. Mutations in this gene are associated with DiGeorge syndrome (DGS), a developmental disorder.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73590

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of a beta-1,4-glucan hydrolase from the snail, Helix pomatia.
J J Marshall et al.
Comparative biochemistry and physiology. B, Comparative biochemistry, 53(2), 231-237 (1976-01-01)
B Grondin et al.
The Journal of biological chemistry, 271(26), 15458-15467 (1996-06-28)
We previously cloned ZNF74, a developmentally expressed zinc finger gene commonly deleted in DiGeorge syndrome. Here, the intron/exon organization of the human gene and the functional properties of the expressed protein are presented. This zinc finger gene from the transcription

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service