Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA003307

Sigma-Aldrich

Anti-TRIM22 antibody produced in rabbit

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-50 kDa-stimulated trans-acting factor antibody produced in rabbit, Anti-RING finger protein 94 antibody produced in rabbit, Anti-Staf-50 antibody produced in rabbit, Anti-Tripartite motif-containing protein 22 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable

immunogen sequence

QVLKELTDVQYYWVDVMLNPGSATSNVAISVDQRQVKTVRTCTFKNSNPCDFSAFGVFGCQYFSSGKYYWEVDVSGKIAWILGVHSKISSLNKRKSSGFAFDPSVNYSKVYSRYRPQYGYWVIGLQNTCEYNAFEDSSSSDPKVLTLFMA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TRIM22(10346)

Looking for similar products? Visit Product Comparison Guide

Immunogen

Tripartite motif-containing protein 22 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TRIM22 (tripartite motif containing 22) gene encodes a protein belonging to the tripartite motif (TRIM) family. It is a ubiquitinated, functional E3 ubiquitin ligase that contains three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Its expression is upregulated by interferon. It mediates antiviral activity against encephalomyocarditis virus, inhibits HIV replication by blocking intracellular trafficking of the viral structural protein Gag to the surface of the cell. The protein ubiquitinates NS5A, a protein essential for hepatitis C virus replication, and inhibits the viral replication.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86656.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yishu Wang et al.
Journal of medical virology, 93(6), 3412-3419 (2020-08-18)
Respiratory syncytial virus (RSV) infection is a major cause of lower respiratory tract disease. Although RSV causes major economic losses every year, effective treatments have not been found so far. Recent studies have shown that the tripartite motif-containing (TRIM) superfamily
Stephen D Barr et al.
PLoS pathogens, 4(2), e1000007-e1000007 (2008-04-05)
Treatment of human cells with Type 1 interferons restricts HIV replication. Here we report that the tripartite motif protein TRIM22 is a key mediator. We used transcriptional profiling to identify cellular genes that were induced by interferon treatment and identified
Patrick Eldin et al.
The Journal of general virology, 90(Pt 3), 536-545 (2009-02-17)
The interferon (IFN) system is a major effector of the innate immunity that allows time for the subsequent establishment of an adaptive immune response against a wide-range of pathogens. Their diverse biological actions are thought to be mediated by the
Chen Yang et al.
Cellular & molecular immunology, 13(1), 94-102 (2015-02-17)
TRIM22, a tripartite-motif (TRIM) protein, is upregulated upon interferon alpha (IFNα) administration to hepatitis C virus (HCV)-infected patients. However, the physiological role of TRIM22 upregulation remains unclear. Here, we describe a potential antiviral function of TRIM22's targeting of the HCV

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service