Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

HPA003263

Sigma-Aldrich

Anti-ZNF143 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SPH-binding factor antibody produced in rabbit, Anti-Zinc finger protein 143 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

TQSGLSQQVTLISQDGTQHVNISQADMQAIGNTITMVTQDGTPITVPAHDAVISSAGTHSVAMVTAEGTEGQQVAIVAQDLAAFHTASSEMGHQQHSHHLVTTETRPLTLVATSNGTQIAVQLGEQPSLEEAIRIASRIQQGET

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF143(7702)

General description

The zinc finger proteins (ZNFs) are made up of anti-parallel hairpin motif. These proteins contain an α-helix, two β-strands and a hairpin structure. The ZNFs are mainly involved in the recognition of DNA, folding and assembly of proteins, transcriptional activation, lipid binding, regulation of apoptosis, and packaging of RNA.

Immunogen

Zinc finger protein 143 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ZNF143 (Zinc finger protein 143) encodes a protein belonging to the kruppel C2H2-type zinc-finger protein family. This protein functions as a transcriptional activator and promotes the transcription of selenocysteine tRNA by binding to SPH motif. It interacts with chromodomain-helicase-DNA binding protein 8 (CHD8) and activates U6 RNA polymerase III transcription .

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84793

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinhong Mei et al.
Acta biochimica et biophysica Sinica, 55(5), 769-782 (2023-05-09)
The clinical oncogenic functions and mechanisms of activating transcription factor 1 (ATF1) in the progression of lung adenocarcinoma have not been completely elucidated. In this study, by employing human lung adenocarcinoma tissues and cells, we detect the correlation of ATF1
Chih-Chi Yuan et al.
Molecular and cellular biology, 27(24), 8729-8738 (2007-10-17)
Chromatin remodeling and histone modification are essential for eukaryotic transcription regulation, but little is known about chromatin-modifying activities acting on RNA polymerase III (Pol III)-transcribed genes. The human U6 small nuclear RNA promoter, located 5' of the transcription start site
J C Rincon et al.
Nucleic acids research, 26(21), 4846-4852 (1998-10-20)
Many vertebrate small nuclear RNA gene promoters contain an SPH motif in their distal control regions that can confer transcriptional stimulation by RNA polymerase II or RNA polymerase III. Using the human U6 gene SPH motif as a probe, we
Lili Zhang et al.
Cancer research, 80(12), 2599-2611 (2020-04-22)
Zinc finger protein 143 (ZNF143) belongs to the zinc finger protein family and possesses transcription factor activity by binding sequence-specific DNA. The exact biological role of ZNF143 in hepatocellular carcinoma (HCC) has not been investigated. Here we report that ZNF143
Identification and Analysis of Disease Target Network of Human MicroRNA and Predicting Promising Leads for ZNF439, a Potential Target for Breast Cancer.
Raghavan V and Manasa D B
International Journal of Bioscience, Biochemistry and Bioinformatics, 2, 358-362 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service