Skip to Content
MilliporeSigma
All Photos(7)

Documents

HPA002859

Sigma-Aldrich

Anti-PRAF2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PRA1 family protein 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

MSEVRLPPLRALDDFVLGSARLAAPDPCDPQRWCHRVINN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRAF2(11230)

Immunogen

PRA1 family protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Prenylated Rab acceptor 1 domain family, member 2 (PRAF2) is a novel 19kDa protein with a prenylated Rab acceptor 1 (PRA1) motif and four transmembrane domains and belongs to the PRAF protein family. The gene is highly expressed in the brain and plays an essential role in neuroblastoma (NB) tumorigenesis metastasis. It may serve as a candidate prognostic marker in NB. Its expression is elevated in various tumors /cancer tissues of the breast, colon, lung and ovary. The PRAF2-related proteins (PRAF1 and PRAF3) play multiple roles in cellular processes (such as endo/exocytic vesicle trafficking and glutamate uptake).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74358

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamás Borsics et al.
Cancer science, 101(7), 1624-1631 (2010-04-24)
Our previous studies revealed that the expression of the 19-kDa protein prenylated Rab acceptor 1 domain family, member 2 (PRAF2) is elevated in cancer tissues of the breast, colon, lung, and ovary, when compared to noncancerous tissues of paired samples.
Dana-Lynn T Koomoa et al.
Neuroscience letters, 436(2), 171-176 (2008-04-09)
PRA1 domain family, member 2 (PRAF2) is a novel 19-kDa protein with a prenylated Rab acceptor 1 (PRA1) motif and four transmembrane domains. Our previous studies revealed that PRAF2 is highly expressed in the brain and serves as a candidate
Lisette P Yco et al.
International journal of oncology, 42(4), 1408-1416 (2013-02-27)
Prenylated Rab acceptor 1 domain family, member 2 (PRAF2) is a novel 19-kDa protein with four transmembrane-spanning domains that belongs to the PRAF protein family. Neuroblastoma (NB) is the most common malignant extracranial solid tumor of childhood that originates in
Zhaoye Qian et al.
BMC cancer, 19(1), 585-585 (2019-06-16)
Prenylated Rab acceptor 1 domain family, member 2 (PRAF2) is involved in the occurrence and progression of several malignant tumors. However, its potential role in esophageal squamous cell carcinoma (ESCC) is still unknown. PRAF2 mRNA expression was determined in 77

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service