Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA001886

Sigma-Aldrich

Anti-IL12A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CLMF p35 antibody produced in rabbit, Anti-Cytotoxic lymphocyte maturation factor 35 kDa subunit antibody produced in rabbit, Anti-IL-12 subunit p35 antibody produced in rabbit, Anti-IL-12A antibody produced in rabbit, Anti-Interleukin-12 subunit α precursor antibody produced in rabbit, Anti-NK cell stimulatory factor chain 1 antibody produced in rabbit, Anti-NKSF1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

RSLLLVATLVLLDHLSLARNLPVATPDPGMFPCLHHSQNLLRAVSNMLQKARQTLEFYPCTSEEIDHEDITKDKTSTVEACLPLELTKNESCLNSRETSFITNGSCLASRKTSFMMALCLSSIYEDLKMYQVEF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL12A(3592)

General description

Interleukin 12 (IL12A) is a 70kDa disulfide-linked heterodimeric cytokine which is associated with various pleiomorphic biological effects on T and NK (natural killer) cells. It is expressed in monocytes/macrophages and B cells. It consists of two covalently linked chains i.e. p40 and p35. It is associated with different biological processes such as inducing cytokine production (e.g. tumor necrosis factor)), IFN (interferon)-γ gene expression, stimulating cytotoxic activity of NK and T cells. It also plays an important role in immunomodulation.

Immunogen

interleukin 12A

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84381

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D'Andrea et al.
The Journal of experimental medicine, 178(3), 1041-1048 (1993-09-01)
Natural killer cell stimulatory factor or interleukin 12 (NKSF/IL-12) is a heterodimeric cytokine produced by monocytes/macrophages, B cells, and possibly other accessory cell types primarily in response to bacteria or bacterial products. NKSF/IL-12 mediates pleiomorphic biological activity on T and
Bhalchandra Mirlekar et al.
Cancer immunology research, 6(9), 1014-1024 (2018-07-08)
Although successes in cancer immunotherapy have generated considerable excitement, this form of treatment has been largely ineffective in patients with pancreatic ductal adenocarcinoma (PDA). Mechanisms that contribute to the poor antitumor immune response in PDA are not well understood. Here
C Yoon et al.
The EMBO journal, 19(14), 3530-3541 (2000-07-19)
Human interleukin-12 (IL-12, p70) is an early pro-inflammatory cytokine, comprising two disulfide-linked subunits, p35 and p40. We solved the crystal structures of monomeric human p40 at 2.5 A and the human p70 complex at 2.8 A resolution, which reveals that
A D'Andrea et al.
The Journal of experimental medicine, 176(5), 1387-1398 (1992-11-01)
Natural killer cell stimulatory factor (NKSF), or interleukin 12 (IL-12), is a 70-kD heterodimeric cytokine composed of two covalently linked chains, p40 and p35. NKSF/IL-12 has multiple effects on T and NK cells and was originally identified and purified from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service