Skip to Content
MilliporeSigma
All Photos(7)

Key Documents

HPA001733

Sigma-Aldrich

Anti-DLG3 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA1232, Anti-MRX90, Anti-NE-Dlg, Anti-NEDLG, Anti-PPP1R82, Anti-SAP-102, Anti-SAP102

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

HKHQHCCKCPECYEVTRLAALRRLEPPGYGDWQVPDPYGPGGGNGASAGYGGYSSQTLPSQAGATPTPRTKAKLIPTGRDVGPVPPKPVPGKSTPKLNGSGPSWWPECTCTNRDWYEQVNGSD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DLG3(1741)

Immunogen

Discs large homolog 3 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In human DLG3 (disks large homolog 3) encodes for synapse-associated protein 102 (SAP102) that belongs to the guanylate kinase protein family. It is found to be expressed during early brain development and is present to the postsynaptic density of excitatory synapses. Structurally, it is composed of three amino-terminal PDZ (PSD95, Dlg1, zo-1) domains, an Src homology domain, and a carboxyl-terminal guanylate kinase domain. The PDZ domains participates in mediating protein-protein interactions and also helps in binding to the short amino acid motifs at the carboxyl termini of interacting proteins. It plays a positive role in clustering of NMDA (N-methyl-D-aspartate) receptors at excitatory synapses as well as it negatively regulate cell proliferation through interaction with the C-terminal region of the polyposis coli tumor suppressor protein. Mutation in DLG3 gene causes a heterogeneous disorder: nonsyndromic X-linked mental retardation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74014

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick Tarpey et al.
American journal of human genetics, 75(2), 318-324 (2004-06-09)
We have identified truncating mutations in the human DLG3 (neuroendocrine dlg) gene in 4 of 329 families with moderate to severe X-linked mental retardation. DLG3 encodes synapse-associated protein 102 (SAP102), a member of the membrane-associated guanylate kinase protein family. Neuronal
Zhongyu Liu et al.
Journal of neuro-oncology, 116(3), 465-476 (2014-01-02)
Glioblastoma multiforme (GBM) is the most malignant glioma. In the current study, 149 astrocytoma gene expression datasets were classified by prediction analysis of microarray. Strikingly, disks large homolog 3 (DLG3), a membrane-associated guanylate kinase-family gene, had the highest score in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service