Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

HPA001598

Sigma-Aldrich

ANTI-ALX1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ALX1, Anti-CART-1, Anti-CART1, Anti-Cartilage homeoprotein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

DNESFYSKASAGKCVQAFGPLPRAEHHVRLERTSPCQDSSVNYGITKVEGQPLHTELNRAMDNCNSLRMSPVKGMQEKGELDELGDKCDSNVSSSKKRRHRTTFTSLQLEELEKVFQKTHYP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ALX1(8092)

Immunogen

Cartilage homeoprotein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ALX1 (ALX homeobox protein 1) gene encodes a protein that functions as a transcriptional activator as well as a repressor. It recognizes palindromic sequence containing two TAAT sequences separated by three or four base pairs. In rodents, it functions in the survival of the forebrain mesenchyme and in chondrocyte differentiation and cervix development. Mutations in this gene cause neural tube defects such as acrania and meroanencephaly. In vertebrates, it is involved in the regulation of skeletal elements of the limbs, face and neck.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST84721

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles A Ettensohn et al.
Development (Cambridge, England), 130(13), 2917-2928 (2003-05-21)
In the sea urchin embryo, the large micromeres and their progeny function as a critical signaling center and execute a complex morphogenetic program. We have identified a new and essential component of the gene network that controls large micromere specification
Q Zhao et al.
Nature genetics, 13(3), 275-283 (1996-07-01)
The paired-class homeobox-containing gene, Cart1, is expressed in forebrain mesenchyme, branchial arches, limb buds and cartilages during embryogenesis. Here, we show that Cart1-homozygous mutant mice are born alive with acrania and meroanencephaly but die soon after birth-a phenotype that strikingly
G Q Zhao et al.
Mechanisms of development, 48(3), 245-254 (1994-12-01)
We have examined the expression of the gene for Cart-1, a new homeodomain-containing protein, during rat embryonic development. In early embryos, Cart-1 RNA was detected at high levels in head mesenchyme, lateral mesoderm, sclerotomes and limb bud mesenchyme. These tissues
R L Cai
Biochemical and biophysical research communications, 250(2), 305-311 (1998-10-01)
Homeodomain proteins play important roles in animal development by controlling the expression of genes involved in determining cell fates. The recently cloned human Cart1 gene encodes a paired-class homeodomain (hCART1), whose rodent homolog is mainly expressed in early chondrocytes and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service