Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HPA001165

Sigma-Aldrich

Anti-KDM6A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-UTX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

SNSCILLDKCPPPRPPTSPYPPLPKDKLNPPTPSIYLENKRDAFFPPLHQFCTNPKNPVTVIRGLAGALKLDLGLFSTKTLVEANNEHMVEVRTQLLQPADENWDPTGTKKIWRCESNRSHTTI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UTY(7404)

Immunogen

lysine (K)-specific demethylase 6A

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Histone demethylase UTY is an enzyme encoded by the UTY gene in humans and is mapped to X-linked homologue at Xp11.2. It consists of six mRNA-transcripts and is a candidate for leukemia therapy development. Transcription of this gene is detected in various human tissues.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74777

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E H Warren et al.
Journal of immunology (Baltimore, Md. : 1950), 164(5), 2807-2814 (2000-02-29)
The mammalian Y chromosome encodes male-specific minor histocompatibility (H-Y) Ags that are recognized by female T cells in an MHC-restricted manner. Two human H-Y epitopes presented by HLA-A2 and HLA-B7, respectively, have been identified previously and both are derived from
Ingeborg Laaser et al.
Omics : a journal of integrative biology, 15(3), 141-154 (2011-02-19)
Over 90% of human genes produce more than one mRNA by alternative splicing (AS). Human UTY (ubiquitously transcribed tetratricopeptide repeat protein on the chromosome Y) has six mRNA-transcripts. UTY is subject to interdisciplinary approaches such as Y chromosomal genetics or
A Greenfield et al.
Human molecular genetics, 7(4), 737-742 (1998-05-16)
We recently have identified a ubiquitously transcribed mouse Y chromosome gene, Uty , which encodes a tetratricopeptide repeat (TPR) protein. A peptide derived from the UTY protein confers H-Y antigenicity on male cells. Here we report the characterization of a
Shahryar E Mir et al.
Oncotarget, 8(45), 78978-78988 (2017-11-08)
Medulloblastoma (MB), the most common malignant childhood brain tumor, encompasses a collection of four clinically and molecularly distinct tumor subgroups, i.e. WNT, SHH, Group 3 and Group 4. These tumors are believed to originate from precursor cells during cerebellar development.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service