Skip to Content
MilliporeSigma
All Photos(1)

Documents

HGECKO2G

Sigma-Aldrich

Gecko2 Human Whole Genome CRISPR Pool, gRNA Only Lenti Particles (Gecko2 vector)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106609
NACRES:
NA.51

packaging

pkg of 8x25 μL (vials)

Quality Level

concentration

5x108  VP/ml (via p24 assay)

application(s)

CRISPR

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

Developed at the Broad Institute by Feng Zhang′s lab, the human GeCKO v2 (two vector system) libraries consist of over 100,000 unique gRNAs for gene knock-out in the human genome. Using a dual lenti CRISPR vector wherein the pooled libraries will only express the gRNA (with the lentiGuide-Puro vector), this system produces over 100X higher titer virus compared to version 1. The lentiGuide-Puro pool should be used only in cell lines with Cas9 already integrated (which can be generated using a separate lenti-Cas9-Blast vector). Sigma′s lentiviral human GeCKO pool guide RNA only is provided in 8 x 25 ul aliquots at a minimum titer of 5X10^8 TU/ml (measured by a p24 assay). Each species-specific library is delivered as two half-libraries (A and B). It is recommended to screen both A and B libraries together, which will include 6 sgRNAs per gene (3 sgRNAs in each library). Both libraries contain 1000 non-targeting control sgRNAs. The A library also targets miRNAs (4 sgRNAs per miRNA).

Application

Functional Genomics/Screening /Target Validation

Features and Benefits

  • Use CRISPR nucleases to knockout protein-coding genes to assess their function
  • Efficiently screen the whole human genome (16,000+ genes) at the bench-top without robotics or specialized equipment
  • Numerous built-in enrichment and depletion controls allow researchers to confidently gauge the success of their pooled screening experiments
  • Lentiviral CRISPRs can infect a broad variety of mammalian cells by transducing a single guide RNA (sgRNA) to a Cas9-expressing human cell line to facilitate gene knockout for screening applications.
  • Use the dual vector system for the human GeCKO version 2 libraries for human cell lines that have Cas9 already integrated into the genome.
  • Use puromycin gRNA selection after transduction.

Preparation Note

Puro Kill Curve and Determining CFU (Colony Formation Unit) per mL. Prior to performing a library-scale screening, two preliminary experiments must be conducted. Visit Sigma.com/pooledscreening.

Other Notes

This product is for R&D use only, not for drug, household, or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices. Though the lentiviral transduction particles produced are replication incompetent, it is recommended that they be treated as Risk Group Level 2 (RGL-2) organisms in laboratory handling. Follow all published RGL-2 guidelines for laboratory handling and waste decontamination.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Articles

Lentiviral vector systems prioritize safety features, with design precautions preventing replication. Good handling practices are essential for use.

Get tips for handling lentiviruses, optimizing experiment setup, titering lentivirus particles, and selecting helpful products for transduction.

Protocols

Lentivirus versions of genome modification technologies support successful CRISPR, RNAi, and ORF experiments.

FACS sorts cells based on light scattering and fluorescence for objective cell analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service