Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HCLPRACDR

Sigma-Aldrich

PR Dynamic Range Analyte ControlDR

Cell line controls for immunohistochemistry and in situ hybridization

Synonym(s):

Progesterone Receptor Dynamic Range Analyte Control

Sign Into View Organizational & Contract Pricing

Select a Size

2 SLIDES
$120.00
5 SLIDES
$219.00
1 EA
$2,340.00

$120.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
2 SLIDES
$120.00
5 SLIDES
$219.00
1 EA
$2,340.00

About This Item

UNSPSC Code:
12352203

$120.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human cell culture

packaging

pkg of 1 ea cell microarray block (HCL034)
pkg of 2 slides (HCL032)
pkg of 5 slides (HCL033)

manufacturer/tradename

Histocyte

storage temp.

2-8°C

General description

The Progesterone Receptor Analyte ControlDR has four cell line cores which demonstrate a range of expression for PR: negative, low/intermediate, intermediate/high, and high.
HistoCyte Laboratories control slides and blocks are made from cultured human cell lines to provide the required range of expression of a specific biomarker. The cell lines are processed using proprietary methods that allow for the creation of a high density embedded cell block with cores that retain their cellular morphology. Available as pre-cut unstained slides or as cell microarray blocks for sectioning.

Application

Ideal for use as a same slide control for immunohistochemistry (IHC) and in situ hybridization (ISH).
The Progesterone Receptor Analyte ControlDR histology slides have four cell line cores providing a dynamic range of expression of PR: negative, low/intermediate, intermediate/high, and high. Analyte controls are placed at one end of slide, allowing sample application on same slide for valid comparison.
The Progesterone Receptor Analyte ControlDR cell microarray block contains four cell line cores that are 2 mm in diameter and between 3.0-3.5 mm in length. The cell microarray block may yield over 300 sections depending on thickness.

Preparation Note

Cell microarray blocks are fixed in 10% neutral buffered formalin and embedded in paraffin wax. Pre-cut unstained slides are prepared from 4 μm thick sections, mounted on positively charged slides, and dried at 37 °C overnight.

Other Notes

For certificates of analysis, contact [email protected].
Progesterone Receptor Analyte ControlDR brochure is available from HistoCyte Laboratories.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service