Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H9279

Sigma-Aldrich

α-Hydroxyfarnesylphosphonic acid

2 mg/mL in ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H27O4P
CAS Number:
Molecular Weight:
302.35
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

concentration

2 mg/mL in ethanol

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C\C(O)P(O)(O)=O

Biochem/physiol Actions

Inhibitor of farnesyltransferase; much weaker activity against geranylgeranyl transferase.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

57.2 °F - closed cup - Solvent

Flash Point(C)

14.0 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J B Gibbs et al.
The Journal of biological chemistry, 268(11), 7617-7620 (1993-04-15)
The ras oncogene product, Ras, is synthesized in vivo as a precursor protein that requires post-translational processing to become biologically active and to be capable of transforming mammalian cells. Farnesylation appears to be a critical modification of Ras, and thus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service