Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H8510

Sigma-Aldrich

Hesperidinase from Penicillium sp.

lyophilized powder, ≥10 units/g solid

Synonym(s):

Hesperidin-α-1,6-rhamnosidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

form

lyophilized powder

specific activity

≥10 units/g solid

storage temp.

2-8°C

Other Notes

Contains both α-L-rhamnosidase (EC 3.2.1.40) and β-D-glucosidase (3.2.1.21) activities.

Unit Definition

One unit will liberate 1.0 μmole of reducing sugar (as glucose) from hesperidin per min at pH 3.8 at 40 °C.

Physical form

Lyophilized powder containing sorbitol

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia Lorena Rojas et al.
Journal of industrial microbiology & biotechnology, 38(9), 1515-1522 (2011-01-12)
Rhamnosidases are enzymes that catalyze the hydrolysis of terminal nonreducing L-rhamnose for the bioconversion of natural or synthetic rhamnosides. They are of great significance in the current biotechnological area, with applications in food and pharmaceutical industrial processes. In this study
L D Varbanets et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 73(5), 9-15 (2011-12-15)
Investigation of 15 different glycoside activities of 64 strains isolated from water and invertebra of the Black Sea has shown that 64% of the studied strains displayed the capacity to synthesize enzymes with alpha-L-ramnosidase activity which varied from 0.01 to
Sudha Chaturvedi et al.
Mycopathologia, 172(4), 247-256 (2011-06-28)
Geomyces destructans is the etiologic agent of bat geomycosis, commonly referred to as white nose syndrome (WNS). This infection has caused severe morbidity and mortality in little brown bats (Myotis lucifugus) and has also spread to other bat species with
E V Gudzenko et al.
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993), 74(2), 14-21 (2012-06-13)
A scheme of isolation and purification of the enzyme with alpha-L-rhamnosidase activity has been developed. It included fractionation by ammonium sulfate and chromatography on TSK-gels Toyopearl HW-60, Fractogel TSK DEAE-650-s and Sepharose 6B. The enzyme was purified 60 times with
Lenka Weignerová et al.
Bioresource technology, 115, 222-227 (2011-09-06)
Extensive screening for a robust producer of α-L-rhamnosidase activity from well-defined strains of filamentous fungi, including multifactorial optimization (inducers, cultivation conditions) was accomplished. Enzyme production of the optimal producer Aspergillus terreus (non-toxigenic) was scaled up to 50L. α-L-Rhamnosidase, which was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service