Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

H7890

Sigma-Aldrich

L-Homoserine lactone hydrochloride

suitable for ligand binding assays

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H7NO2 · HCl
CAS Number:
Molecular Weight:
137.56
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Homoserine lactone hydrochloride,

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

Cl.N[C@H]1CCOC1=O

InChI

1S/C4H7NO2.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1

InChI key

XBKCXPRYTLOQKS-DFWYDOINSA-N

General description

L-Homoserine lactone (HSL), also called acyl-HSL, comprises a homoserine lactone ring and a fatty acyl side.[1] The levels of acyl-HSL in bacteria is dictated by the availability of the substrates and acyl-homoserine lactones synthase.[2]

Application

L-Homoserine lactone hydrochloride has been used as an inhibitor for arterial smooth muscle contraction.[3]

Biochem/physiol Actions

L-Homoserine lactone inhibition reduces virulence factors and halts inflammation and tissue damage.[1] In Pseudomonas aeruginosa it controls virulence factor production and biofilm formation. In Agrobacterium tumefaciens, it is essential for conjugal transfer.[4]
Acylated L-Homoserine lactone(s) are used to study bacterial quorum-sensing signaling mechanisms.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long-chain acyl-homoserine lactone quorum-sensing regulation of Rhodobacter capsulatus gene transfer agent production
Schaefer AL, et al.
Journal of Bacteriology, 184(23), 6515-6521 (2002)
Defining the structure and function of acyl-homoserine lactone autoinducers
Churchill MEA, et al.
Methods in Molecular Biology, 692(23), 159-171 (2011)
The Pseudomonas aeruginosa quorum-sensing signal molecule, N-(3-oxododecanoyl)-l-homoserine lactone, inhibits porcine arterial smooth muscle contraction
Lawrence RN, et al.
British Journal of Pharmacology, 128(4), 845-848 (1999)
Inhibiting N-acyl-homoserine lactone synthesis and quenching Pseudomonas quinolone quorum sensing to attenuate virulence
Chan KG, et al.
Frontiers in Microbiology, 6, 1173-1173 (2015)
Adam Schikora et al.
Plant physiology, 157(3), 1407-1418 (2011-09-24)
Pathogenic and symbiotic bacteria rely on quorum sensing to coordinate the collective behavior during the interactions with their eukaryotic hosts. Many Gram-negative bacteria use N-acyl-homoserine lactones (AHLs) as signals in such communication. Here we show that plants have evolved means

Questions

  1. How to dissolve L-Homoserine LA tone hydrochloride

    1 answer
    1. The solubility of this product is tested at 50 mg/mL in water. Please see the link below to review a sample Certificate of Analysis:
      https://www.sigmaaldrich.com/certificates/COFA/H7/H7890/H7890-BULK________SLCG7532__.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service