Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H7395

Sigma-Aldrich

Hepatitis B virus pre-S region fragment 120-145

≥95% (HPLC)

Synonym(s):

Met-Gln-Trp-Asn-Ser-Thr-Thr-Phe-His-Gln-Thr-Leu-Gln-Asp-Pro-Arg-Val-Arg-Gly-Leu-Tyr-Phe-Pro-Ala-Gly-Gly

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C135H199N39O38S
CAS Number:
Molecular Weight:
3008.33
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

storage temp.

−20°C

SMILES string

CSCC[C@H](N)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](Cc1c[nH]c2ccccc12)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CO)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H](Cc4c[nH]cn4)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CC(O)=O)C(=O)N5CCC[C@H]5C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CCCNC(N)=N)C(=O)NCC(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc6ccc(O)cc6)C(=O)N[C@@H](Cc7ccccc7)C(=O)N8CCC[C@H]8C(=O)N[C@@H](C)C(=O)NCC(=O)NCC(O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A R Neurath et al.
Vaccine, 4(1), 35-37 (1986-03-01)
Studies with synthetic peptides have provided evidence for the presence of preS coded sequences in the envelope (env) proteins of hepatitis B virus (HBV) and indicated that these sequences are involved in the specific attachment of HBV to liver cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service