Skip to Content
MilliporeSigma
All Photos(1)

Documents

H6894

Sigma-Aldrich

[Tyr(SO3H)63]-Hirudin Fragment 54-65

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H93N13O28S
CAS Number:
Molecular Weight:
1548.58
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

biological source

synthetic (organic)

Quality Level

Assay

≥95% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(Cc1ccccc1)NC(=O)C(CC(O)=O)NC(=O)CN)C(=O)N2CCCC2C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(Cc3ccc(OS(O)(=O)=O)cc3)C(=O)NC(CC(C)C)C(=O)NC(CCC(N)=O)C(O)=O

InChI

1S/C66H93N13O28S/c1-5-34(4)55(78-59(95)41(21-26-53(88)89)71-56(92)38(18-23-50(82)83)72-61(97)44(29-35-10-7-6-8-11-35)77-63(99)46(31-54(90)91)69-49(81)32-67)65(101)79-27-9-12-47(79)64(100)73-40(20-25-52(86)87)57(93)70-39(19-24-51(84)85)58(94)76-45(30-36-13-15-37(16-14-36)107-108(104,105)106)62(98)75-43(28-33(2)3)60(96)74-42(66(102)103)17-22-48(68)80/h6-8,10-11,13-16,33-34,38-47,55H,5,9,12,17-32,67H2,1-4H3,(H2,68,80)(H,69,81)(H,70,93)(H,71,92)(H,72,97)(H,73,100)(H,74,96)(H,75,98)(H,76,94)(H,77,99)(H,78,95)(H,82,83)(H,84,85)(H,86,87)(H,88,89)(H,90,91)(H,102,103)(H,104,105,106)

InChI key

CKXMQSIWBKKXGT-UHFFFAOYSA-N

Amino Acid Sequence

Gly-Asp-Phe-Glu-Glu-Ile-Pro-Glu-Glu-Tyr(SO3H)-Leu-Gln

Application

[Tyr(SO3H)63]-hirudin fragment 54-65 has been used to study the effect of thrombin in presence of leupeptin. It is also used to study the effect of hirudin peptide on the thrombin-rHCII (recombinant heparin cofactor II)-L444R (leucine 444 to arginine) complex separation.

Biochem/physiol Actions

Hirudin fragment that binds to the regulatory exosite I on thrombin and the regulatory proexosite I on prothrombin. The interaction with thrombin prevents the thrombin-induced activation of clotting factor V. The interaction with prothrombin inhibits the formation of thrombin by factor Xa in the presence of factor Va.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Akiba et al.
European journal of biochemistry, 259(3), 643-650 (1999-03-27)
We investigated the regulation of arachidonic acid liberation catalyzed by group-IV cytosolic phospholipase A2 (cPLA2) in human platelets upon stimulation with thrombin through interaction with protease-activated receptor-1 (PAR-1) or glycoprotein Ib. Leupeptin, a protease inhibitor, completely inhibited thrombin-induced arachidonic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service