Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6791

Sigma-Aldrich

LEFTY-B human

HumanKine®, >90% (SDS-PAGE), recombinant, expressed in HEK 293 cells, lyophilized powder, suitable for cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$403.00

$403.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
$403.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$403.00


Please contact Customer Service for Availability

Product Name

LEFTY-B human, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture

biological source

human

Quality Level

recombinant

expressed in HEK 293 cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

≤2.5 ng/mL EC50

mol wt

monomer 38 kDa (glycosylated)

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... LFTY1(10637)

Physical form

Lyophilized from a 0.2 μm filtered solution of 10 mM Tris-HCl pH 7.4 + 50 mM NaCl + 0.5% CHAPS.

Analysis Note

The specific activity was determined by dose dependent ability to inhibit BMP-4 (6.5 ng/mL) induction of alkaline phosphatase production in the MC-3T3-E1 cell line (mouse chondrogenic cell line).

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service