Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H6769

Sigma-Aldrich

Hirudin Fragment 54-65 non-sulfated

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H93N13O25
CAS Number:
Molecular Weight:
1468.52
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CCC(O)=O)NC(=O)C(CCC(O)=O)NC(=O)C(Cc1ccccc1)NC(=O)C(CC(O)=O)NC(=O)CN)C(=O)N2CCCC2C(=O)NC(CCC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(Cc3ccc(O)cc3)C(=O)NC(CC(C)C)C(=O)NC(CCC(N)=O)C(O)=O

Amino Acid Sequence

Gly-Asp-Phe-Glu-Glu-Ile-Pro-Glu-Glu-Tyr-Leu-Gln

Biochem/physiol Actions

Hirudin fragment that binds to the regulatory exosite I (proexosite I) on prothrombin with lower affinity than the sulfated peptide. It inhibits the activation of prothrombin by factor Xa in the presence of factor Va.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service