Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H5636

SAFC

3β-Hydroxychol-5-en-24-oic acid 3-sulfate sodium salt

Synonym(s):

3β-Hydroxy-5-cholen-24-oic acid 3-sulfate, 5-Cholen-24-oic acid-3β-ol-3-sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C24H36O6SNa2
CAS Number:
Molecular Weight:
498.58
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parmentier, G. and Eyssen, H.
Journal of Chromatography A, 152, 285-285 (1978)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service