Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H4521

Sigma-Aldrich

HIV Envelope protein (gp41) fragment 579-601

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C119H196N34O32S
CAS Number:
Molecular Weight:
2647.10
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)CNC(=O)[C@H](CC(C)C)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1ccc(O)cc1)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](N)CCCNC(N)=N)[C@@H](C)CC)C(C)C)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)NCC(=O)N[C@@H](CS)C(=O)N[C@@H](CO)C(=O)NCC(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C119H196N34O32S/c1-16-64(13)96(115(182)149-84(51-68-53-131-72-28-19-18-26-70(68)72)101(168)132-55-91(159)137-87(58-186)113(180)150-86(57-154)102(169)134-54-90(158)136-78(117(184)185)30-21-23-43-121)151-92(160)56-133-100(167)79(46-59(3)4)143-110(177)82(49-62(9)10)144-107(174)76(37-40-89(124)157)140-105(172)75(36-39-88(123)156)141-112(179)85(52-94(163)164)147-103(170)73(29-20-22-42-120)139-109(176)81(48-61(7)8)145-111(178)83(50-67-32-34-69(155)35-33-67)146-104(171)74(31-25-45-130-119(127)128)138-106(173)77(38-41-93(161)162)142-114(181)95(63(11)12)152-98(165)66(15)135-108(175)80(47-60(5)6)148-116(183)97(65(14)17-2)153-99(166)71(122)27-24-44-129-118(125)126/h18-19,26,28,32-35,53,59-66,71,73-87,95-97,131,154-155,186H,16-17,20-25,27,29-31,36-52,54-58,120-122H2,1-15H3,(H2,123,156)(H2,124,157)(H,132,168)(H,133,167)(H,134,169)(H,135,175)(H,136,158)(H,137,159)(H,138,173)(H,139,176)(H,140,172)(H,141,179)(H,142,181)(H,143,177)(H,144,174)(H,145,178)(H,146,171)(H,147,170)(H,148,183)(H,149,182)(H,150,180)(H,151,160)(H,152,165)(H,153,166)(H,161,162)(H,163,164)(H,184,185)(H4,125,126,129)(H4,127,128,130)/t64-,65-,66-,71-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,95-,96-,97-/m0/s1

InChI key

PBKBSKTYOPFGSZ-HEQQLXNASA-N

Amino Acid Sequence

Arg-Ile-Leu-Ala-Val-Glu-Arg-Tyr-Leu-Lys-Asp-Gln-Gln-Leu-Leu-Gly-Ile-Trp-Gly-Cys-Ser-Gly-Lys

General description

Peptide corresponding to the region of the HIV envelope protein (gp41) identified as a major epitope recognized by antibodies of AIDS patients.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Kemp et al.
Science (New York, N.Y.), 241(4871), 1352-1354 (1988-09-09)
An antibody detection procedure based on agglutination of autologous red cells has been developed for samples of whole blood. A nonagglutinating monoclonal antibody to human red blood cells conjugated to a synthetic peptide antigen (in this case residues 579 to
J J Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 83(16), 6159-6163 (1986-08-01)
A peptide of 21 amino acids with the sequence Arg-Ile-Leu-Ala-Val-Glu-Arg-Tyr-Leu-Lys-Asp-Gln-Gln-Leu-Leu-Gly-Ile-Trp- Gly-Cys - Ser encoded by a segment in the env gene of human T-lymphotropic virus type III (HTLV-III), corresponding to amino acids 586-606 of the precursor envelope glycoprotein, has been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service