Skip to Content
MilliporeSigma
All Photos(1)

Documents

H2791

Sigma-Aldrich

Anti-HOXB8 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Hox-2.4, Anti-Hox-2D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~30 kDa

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

western blot: 1-2 μg/mL using Jurkat cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOXB8(3218)

General description

Homeobox B8 (HOXB8) protein belongs to the homeobox (HOX) containing complex. HOXB8 gene is expressed in the spinal cord during embryonic development. The HOXB8 gene is located on the human chromosome at 17q21.32.

Specificity

Anti-HOXB8 (C-terminal) recognizes human HOXB8 (also known as Hox-2D, Hox-2).

Application

Anti-HOXB8 (C-terminal) antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Overexpression of the homeobox B8 (HOXB8) gene is identified as a cause for leukemia. The encoded protein prevents myeloid differentiation by a mechanism that requires interaction with pre‐B‐cell leukemia homeobox (Pbx) protein.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

Store at –20 °C. For continuous use, the product may be stored at 2–8 °C for up to one month. For extended storage, freeze in working aliquots at –20 °C. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan C Holstege et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(17), 6338-6343 (2008-04-24)
Although Hox gene expression has been linked to motoneuron identity, a role of these genes in development of the spinal sensory system remained undocumented. Hoxb genes are expressed at high levels in the dorsal horn of the spinal cord. Hoxb8
P S Knoepfler et al.
Oncogene, 20(39), 5440-5448 (2001-09-26)
HoxB8 was the first homeobox gene identified as a cause of leukemia. In murine WEHI3B acute myeloid leukemia (AML) cells, proviral integration leads to the expression of both HoxB8 and Interleukin (IL-3). Enforced expression of HoxB8 blocks differentiation of factor-dependent
Joy M Greer et al.
Neuron, 33(1), 23-34 (2002-01-10)
Repertoires of grooming behaviors critical to survival are exhibited by most animal species, including humans. Genes that influence this complex behavior are unknown. We report that mice with disruptions of Hoxb8 show, with 100% penetrance, excessive grooming leading to hair
Nobumoto Tomioka et al.
Cancer genetics and cytogenetics, 201(1), 6-14 (2010-07-17)
Unlike the case with some other solid tumors, whole genome array screening has not revealed prognostic genetic aberrations in primary gastric cancer. Comparative genomic hybridization (CGH) using bacterial artificial chromosome (BAC) arrays for 56 primary gastric cancers resulted in identification

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service