Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H2634

Sigma-Aldrich

Helveticoside

Synonym(s):

Alleoside A, Deglucoerysimoside, Erisimin, Erysimotoxin, Strophanthidin 3β-digitoxoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42O9
CAS Number:
Molecular Weight:
534.64
EC Number:
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

storage temp.

2-8°C

SMILES string

O[C@@]12[C@]3([H])CC[C@]4(O)C[C@@H](O[C@]5([H])O[C@H](C)[C@@H](O)[C@@H](O)C5)CC[C@]4(C([H])=O)[C@@]3([H])CC[C@]1(C)[C@@H](C(CO6)=CC6=O)CC2

InChI

1S/C29H42O9/c1-16-25(33)22(31)12-24(37-16)38-18-3-8-27(15-30)20-4-7-26(2)19(17-11-23(32)36-14-17)6-10-29(26,35)21(20)5-9-28(27,34)13-18/h11,15-16,18-22,24-25,31,33-35H,3-10,12-14H2,1-2H3/t16-,18+,19-,20+,21-,22+,24+,25-,26-,27+,28+,29+/m1/s1

InChI key

QBILRDAMJUPXCX-AGAUEGNUSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service