Skip to Content
MilliporeSigma
All Photos(1)

Documents

H2542

Sigma-Aldrich

Histone H3.3 human

Synonym(s):

Histone, Human

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.25

biological source

human

Quality Level

recombinant

expressed in E. coli

form

liquid

concentration

1 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... H3-3A(3020)

Looking for similar products? Visit Product Comparison Guide

Other Notes

Supplied as 1 mg/mL in 20 mM NaPO4, pH 7.0, 0.3 M NaCl, 1 mM EDTA, and 1 mM DTT.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service