Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H2029

Sigma-Aldrich

Herpes simplex virus-I proteinase substrate II

≥90% (HPLC)

Synonym(s):

Gly-Ile-Ala-Gly-His-Thr-Tyr-Leu-Gln-Ala-Ser-Glu-Lys-Phe-Lys-Nle-Trp-Gly-Ala-Glu

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C102H152N26O29
CAS Number:
Molecular Weight:
2206.46
MDL number:
UNSPSC Code:
12352204

Assay

≥90% (HPLC)

composition

Peptide content, ~60%

storage temp.

−20°C

SMILES string

CCCC[C@H](NC(=O)[C@H](CCCCN)NC(=O)[C@H](Cc1ccccc1)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CO)NC(=O)[C@H](C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc2ccc(O)cc2)NC(=O)[C@@H](NC(=O)[C@H](Cc3c[nH]cn3)NC(=O)CNC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)CN)[C@@H](C)CC)[C@@H](C)O)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)NCC(=O)N[C@@H](C)C(=O)N[C@@H](CCC(O)=O)C(O)=O

Other Notes

Contains the putative cleavage site essential for HSV-I proteinase maturational release.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Y Liu et al.
Journal of virology, 65(1), 206-212 (1991-01-01)
The herpes simplex virus 1 (HSV-1) genome specifies an abundant capsid protein which in denaturing gels forms multiple bands designated family 35 proteins (D.K. Braun, B. Roizman, and L. Pereira, J. Virol. 49:142-153, 1984). Nucleotide-sequencing studies have assigned the coding
A R Welch et al.
Journal of virology, 65(8), 4091-4100 (1991-08-01)
The genomic region encoding the assembly protein of simian cytomegalovirus (CMV) strain Colburn has been cloned, sequenced, and found to be organized as a nested set of four in-frame, 3'-coterminal genes, each with its own TATA promoter element and translational
A R Welch et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(23), 10792-10796 (1991-12-01)
A herpesvirus proteinase activity has been identified and partially characterized by using the cloned enzyme and substrate genes in transient transfection assays. Evidence is presented that the proteinase gene of cytomegalovirus strain Colburn encodes a 590-amino acid protein whose N-terminal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service