Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

H0885

Sigma-Aldrich

N-Hexadecyl-L-hydroxyproline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H41NO3
CAS Number:
Molecular Weight:
355.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

suitability

suitable for HPLC stationary phase for resolution of racemic α-amino acids

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCN1C[C@H](O)C[C@H]1C(O)=O

InChI

1S/C21H41NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-22-18-19(23)17-20(22)21(24)25/h19-20,23H,2-18H2,1H3,(H,24,25)/t19-,20+/m1/s1

InChI key

FDSLJAYOPNMZIY-UXHICEINSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Devankov, V.A., et al.
Chromatographia, 13, 677-677 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service