Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW22823

Sigma-Aldrich

Anti-BAT3 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, rat

manufacturer/tradename

Genway 15-288-22823

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAT3(7917)

General description

BAT3 is a gene from major histocompatibility complex. In humans, it is localized in the class III region of the human major histocompatibility complex on chromosome 6. It consists of an ubiquitin homology region at the N-terminal end, short tracts of polyproline and polyglycine, a zinc finger-like domain and a nuclear localization signal (NLS) at the C- terminus end. It is a member of BAG family of proteins. The major responsibility of BAG proteins is to negatively control the chaperone function of the Hsp70 family.

Immunogen

Immunogen Sequence: GI # 18375632, sequence 2-250
Recombinant HLA-B associated transcript-3 isoform b

Application

Anti-BAT3 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Large proline-rich protein BAT3 is a protein encoded by the BAT3 gene in humans. It is very rich in proline and includes short tracts of polyproline, polyglycine and charged amino acids. BAT3 is similar to other proteins with large proline-rich domains, such as some nuclear proteins, collagens, elastin and synapsin. It has distinct functions in the cytosol and in the nucleus. It is also associated with controlling of apoptosis and regulating heat shock protein (HSP) 70 activity.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S T Manchen et al.
Biochemical and biophysical research communications, 287(5), 1075-1082 (2001-10-06)
Human Scythe (also known as BAT3) has been implicated in the control of apoptosis and regulating heat shock protein (HSP) 70 activity. We have attempted to further characterize the role of human Scythe in HeLa cells by studying the cellular
Manu Rangachari et al.
Nature medicine, 18(9), 1394-1400 (2012-08-07)
T cell immunoglobulin and mucin domain–containing 3 (Tim-3) is an inhibitory receptor that is expressed on exhausted T cells during infection with HIV-1 and hepatitis C virus. By contrast, Tim-3 expression and function are defective in multiple human autoimmune diseases.
Toru Sasaki et al.
Genes & development, 21(7), 848-861 (2007-04-04)
In response to DNA damage, p53 undergoes post-translational modifications (including acetylation) that are critical for its transcriptional activity. However, the mechanism by which p53 acetylation is regulated is still unclear. Here, we describe an essential role for HLA-B-associated transcript 3
Nadine Kämper et al.
PloS one, 7(4), e35972-e35972 (2012-05-05)
The human lymphocyte antigen (HLA) encoded BAT3/BAG6 recently attracted interest as a regulator of protein targeting and degradation, a function that could be exerted in the cytosol and in the nucleus. The BAT3 gene was described to consist of 25
J Banerji et al.
Proceedings of the National Academy of Sciences of the United States of America, 87(6), 2374-2378 (1990-03-01)
A large number of genes has been identified previously between the class I and class II gene families within the class III region of the human major histocompatibility complex. The complete sequences of two of these genes, BAT2 and BAT3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service