Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW21286A

Sigma-Aldrich

Anti-EWSR1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RNA-binding protein EWS

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-21286A

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EWSR1(2130)

Immunogen

Immunogen Sequence: GI # 4885225, sequence 380-448
Recombinant ewing sarcoma breakpoint region 1 isoform EWS

Application

Anti-EWSR1 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

RNA-binding protein EWS is a protein encoded by the EWSR1 gene in humans and is located on human chromosome 22. It is involved in a wide variety of sarcomas as well as some non-mesenchymal tumours. EWSR1 is a ′promiscuous′ gene that can fuse with many different partner genes, resulting in identical tumours. It can partner with the same genes in morphologically and behaviourally different neoplasms. EWSR1 rearrangement is also associated with a variety of salivary gland lesions including classic myoepitheliomas and hyalinizing clear cell carcinomas (HCCCs). It also acts as a novel modulator of cis-acting replication element (CRE) function in hepatitis C virus (HCV) replication.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Delattre et al.
Nature, 359(6391), 162-165 (1992-09-10)
Ewing's sarcoma and related subtypes of primitive neuroectodermal tumours share a recurrent and specific t(11;22) (q24;q12) chromosome translocation, the breakpoints of which have recently been cloned. Phylogenetically conserved restriction fragments in the vicinity of EWSR1 and EWSR2, the genomic regions
Cyril Fisher
Histopathology, 64(1), 134-150 (2013-12-11)
Many soft tissue sarcomas have chromosomal translocations with resultant formation of new fusion genes. Among the genes that can be rearranged, the EWSR1 gene has been identified as a partner in a wide variety of clinically and pathologically diverse sarcomas
Todd E Oakland et al.
Journal of virology, 87(12), 6625-6634 (2013-04-05)
The hepatitis C virus (HCV) genome contains numerous RNA elements that are required for its replication. Most of the identified RNA structures are located within the 5' and 3' untranslated regions (UTRs). One prominent RNA structure, termed the cis-acting replication
Akeesha A Shah et al.
The American journal of surgical pathology, 37(4), 571-578 (2013-02-22)
The Ewing sarcoma breakpoint region 1 (EWSR1) is translocated in many sarcomas. Recently, its rearrangement has been described in salivary gland hyalinizing clear cell carcinomas (HCCCs) and in a subset of soft tissue myoepitheliomas. This study examines the presence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service