Skip to Content
MilliporeSigma
All Photos(1)

Documents

GW21258

Sigma-Aldrich

Anti-PPARγ antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Peroxisome proliferator-activated receptor γ

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human, mouse

manufacturer/tradename

Genway 15-288-21258

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPARG(5468)

Immunogen

Immunogen Sequence: GI # 4826930, sequence 1-110
Recombinant peroxisome proliferative activated receptor gamma, isoform 1

Application

Anti-PPARγ antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Peroxisome proliferator-activated receptor γ(PPAR-γ or PPARG) is a type II nuclear receptor encoded by the PPARG gene in humans. PPARs are ligand-activated transcription factors of the nuclear hormone receptor superfamily. PPARγ is expressed as two isoforms, PPARγ2 and PPARγ1. PPARγ2 is expressed at high levels in the adipose tissues, whereas PPARγ1 has a broader expression pattern. PPARγ2 in skeletal muscle can promote conversion of subcutaneous fat to brown fat formation leading to increased energy metabolisms and combating obesity. PPARγ mRNA is expressed in humans in spleen, bone marrow, liver, testis, skeletal muscle and brain, in addition to fat.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Zhou et al.
Biochemical and biophysical research communications, 446(4), 959-964 (2014-03-20)
Brown adipose tissue (BAT) is specialized to dissipate energy as heat, therefore reducing fat deposition and counteracting obesity. Brown adipocytes arise from myoblastic progenitors during embryonic development by the action of transcription regulator PRDM16 binding to PPARγ, which promotes BAT-like
Jaakko L O Pohjoismäki et al.
Nucleic acids research, 40(14), 6595-6607 (2012-04-18)
Cardiomyocyte development in mammals is characterized by a transition from hyperplastic to hypertrophic growth soon after birth. The rise of cardiomyocyte cell mass in postnatal life goes along with a proportionally bigger increase in the mitochondrial mass in response to
Juan Pablo Lezana et al.
Developmental neurobiology, 76(6), 688-701 (2015-10-09)
PPARγ is a ligand-activated nuclear receptor best known for its involvement in adipogenesis and glucose homeostasis. PPARγ activity has also been associated with neuroprotection in different neurological disorders, but the mechanisms involved in PPARγ effects in the nervous system are
Liliane Michalik et al.
Pharmacological reviews, 58(4), 726-741 (2006-11-30)
The three peroxisome proliferator-activated receptors (PPARs) are ligand-activated transcription factors of the nuclear hormone receptor superfamily. They share a high degree of structural homology with all members of the superfamily, particularly in the DNA-binding domain and ligand- and cofactor-binding domain.
A Elbrecht et al.
Biochemical and biophysical research communications, 224(2), 431-437 (1996-07-16)
We describe the molecular cloning and expression of cDNAs encoding human PPAR gamma 1 and PPAR gamma 2. Our sequences are distinct from the published sequence at 3 positions, resulting in nonconservative amino acid substitutions. In humans, PPAR gamma mRNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service