Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GW20021F

Sigma-Aldrich

Anti-Fibronectin Antibody

chicken polyclonal

Synonym(s):

Anti-Cold-insoluble globulin, Anti-FN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

product name

Anti-Fibronectin antibody produced in chicken, affinity isolated antibody, buffered aqueous solution

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-20021F

concentration

1.0-1.2 mg/mL

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN1(2335)

General description

The gene FN1 (Fibronectin) is mapped to human chromosome 2q34. It is present as a soluble dimeric form in plasma. At the cell surface and in the extracellular matrix, it is present as a dimeric or multimeric form.

Immunogen

Fibronectin native protein.
Immunogen Sequence: GI # 31397, sequence 1-2328

Application

Anti-Fibronectin antibody produced in chicken is suitable for western blotting at a working dilution of 1:500 and for cell staining at a working dilution of 1:200. It is also suitable for indirect ELISA.

Biochem/physiol Actions

Fibronectin (FN1) is a glycoprotein that is involved in cell adhesion and migration processes, such as embryogenesis, wound healing, blood coagulation, host defense, and metastasis. It binds to cell surfaces and compounds, such as collagen, fibrin, heparin, fibulin, integrins, cell adhesive mac-2 binding protein (M2BP), cartilage oligomeric matrix protein (COMP), DNA, and actin. Fibronectin interaction with FLRG (Follistatin-related gene protein) and follistatin is important for human hematopoietic cell adhesiveness. It is associated with glomerulopathy with fibronectin deposits, an autosomal dominant disease. Polymorphism in gene is linked with increased risk of knee osteoarthritis. Human pathogens, Staphylococcus aureus and Streptococcus pyogenes, target fibronectin for adhesion to and invasion of host cells. Pigment epithelium-derived factor (PEDF) down-regulates fibronectin to suppress breast cancer metastasis.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Ffrench-Constant et al.
The Journal of cell biology, 109(2), 903-914 (1989-08-01)
The adhesive extracellular matrix glycoprotein fibronectin (FN) is thought to play an important role in the cell migration associated with wound healing. Immunolocalization studies show abundant FN in healing wounds; however, these studies cannot define the cellular site(s) of FN
E Engvall et al.
International journal of cancer, 20(1), 1-5 (1977-07-15)
Fibronectin, a plasma protein immunologically identical with a major surface protein of normal fibroblasts, was found to bind to collagen and gelatin. A solid phase enzyme immunoassay was used for the binding tests. Collagen, gelatin or various control proteins were
R Carlsson et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(4), 2403-2406 (1981-04-01)
Laminin, a basement membrane glycoprotein isolated from cultures of mouse endodermal cells and rat yolk sac carcinoma cells, promoted the attachment of liver cells obtained from regenerating mouse liver. Cells from normal mouse liver attached readily to dishes coated with
Mohamed Reda Fazazi et al.
Nature communications, 15(1), 5404-5404 (2024-06-27)
B cells and T cells collaborate in multiple sclerosis (MS) pathogenesis. IgH[MOG] mice possess a B cell repertoire skewed to recognize myelin oligodendrocyte glycoprotein (MOG). Here, we show that upon immunization with the T cell-obligate autoantigen, MOG[35-55], IgH[MOG] mice develop
Honghai Hong et al.
Breast cancer research and treatment, 148(1), 61-72 (2014-10-07)
Pigment epithelium-derived factor (PEDF) plays an important role in the tumor growth and metastasis inhibition. It has been reported that PEDF expression is significantly reduced in breast cancer, and associated with disease progression and poor patient outcome. However, the exact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service