Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

GN772288

Greiner filter tips

universal, volume 100 μL, 50 mm, natural, sterile, racked

Synonym(s):

Greiner filter tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
packaging:
pack of 960 ea
manufacturer/tradename:
Greiner 772288

packaging

pack of 960 ea

manufacturer/tradename

Greiner 772288

availability

available only in China

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Pipette tips with filter inserts prevent contamination with liquids and aerosols during the pipetting process.
In combination with conventional pipettors, Greiner filter tips prevent the aspiration of particles into the interior of the pipettor. This minimises the danger of unwanted cross-contaimination. The special ultra-micro filter made of hydrophobic polyethylene has a pore size of approx. 10 microns and acts as a reliable barrier against the transfer of aerosols into the shaft of the pipettor.
  • Reccomended for work with DNA and RNA and when handling radioactive materials
  • Maintain the tolerane limits stipulated for pipette tips in DIN 142650.
  • DNase, RNase and human DNA free

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service