Key Documents
GN650160
Greiner CELLSTAR® multiwell culture plates
96 well, TC treated, clear
Synonym(s):
culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates
About This Item
UNSPSC Code:
41122107
material:
polystyrene
Pricing and availability is not currently available.
material
polystyrene
sterility
sterile
packaging
pack of 100 ea
manufacturer/tradename
Greiner 650160
availability
available only in China
General description
The chimney shape design, in combination with the condensation rings in the lid, effectively reduces the risk of cross-contamination and reduces evaporation from the wells. All plates can be stacked on top of each other, both with and without lids. The special design of the plates enables rapid temperature exchange across the entire base area of ​​each individual plate, even when the plates are stacked on top of each other. The standardization of depth, diameter and arrangement of the wells simplifies manual and automated work, as well as the use of common membrane insert systems. All plates have a printed lot number and are both pyrogen-free and DNase/RNase-free.
Legal Information
CELLSTAR is a registered trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)