Key Documents
GN445835
Greiner 3-D Amino HTA plate
non-sterile, 1 test per plate, 25 ea
About This Item
UNSPSC Code:
41121800
packaging
pack of 25 ea
manufacturer/tradename
Greiner 445835
availability
available only in China
suitability
suitable for (cell culture)
General description
The HTAmultifunctional High-Throughput microArraying in Slide Format. 3-D Amino surface meets the needs of DNA, protein and tissue immobilisation. The HTA 1 can be used for the processing of one sample. The HTA 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0.5 mm. Twelve samples can be processed simultaneously. Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Legal Information
HTA is a trademark of Greiner Bio-One GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)