Skip to Content
MilliporeSigma
All Photos(1)

Documents

GEUS78210

illustra ExoProStar

Cytiva US78210, sufficient for 100 reactions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.52

usage

sufficient for 100 reactions

manufacturer/tradename

Cytiva US78210

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Enzymatic PCR clean up technology with illustra™ Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes.illustra™ ExoProStar is a new name for this PCR clean-up technology. No change in the formulation, so you get the same great product, and same great benefits! The ExoProStar PCR and Sequence Reaction Clean-Up Kit updates existing enzymatic PCR clean up technology with illustra™ Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. illustra™ ExoProStar improves digestion efficiency with no degradation of the target PCR product. Your sample is completely intact and ready for immediate use in manual or automated processes.

Features and Benefits

  • Enzymes optimized to work together for high efficiency removal of unincorporated primers and nucleotides
  • Enzymes provided in two separate tubes; just two simple pipetting steps are needed to prepare the reaction
  • Fast 30 min protocol
  • Scalable for different reaction sizes
  • No loss of PCR product
  • Easy to automate
  • Complete heat inactivation of the enzymes within 15 min

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

ExoProStar is a trademark of Cytiva
illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular phylogeny of trigonostomine turbellarians (Platyhelminthes: Rhabdocoela: Trigonostomidae), including four new species from the Northeast Pacific Ocean.
Van S, et al.
Zoological journal of the Linnean Society, 182(2), 237-257 (2017)
Complete Coding Genome Sequences of Uncommon GII. 8 Sapovirus Strains Identified in Diarrhea Samples Collected from Peruvian Children.
Tsinda E K, et al.
Genome Announcements, 5(43), e01137-e01117 (2017)
Horizontal transmission and phylogenetic analysis of bovine leukemia virus in two districts of Miyazaki, Japan.
Mekata H, et al.
The Journal of Veterinary Medical Science, 77(9), 1115-1120 (2015)
Maria Pujantell et al.
Scientific reports, 9(1), 19848-19848 (2019-12-29)
Infection by human papillomavirus (HPV) alters the microenvironment of keratinocytes as a mechanism to evade the immune system. A-to-I editing by ADAR1 has been reported to regulate innate immunity in response to viral infections. Here, we evaluated the role of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service